Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01444130_g1
          See other NEWGENE_1596188 GE Assays ›
          Gene Symbol:
          NEWGENE_1596188 Pigz
          Gene Name:
          nuclear cap binding protein subunit 2, 20kDa
          phosphatidylinositol glycan anchor biosynthesis, class Z
          Gene Aliases:
          Ncbp2
          Ncbp2
          Chromosome Location:
          Chr.11: 72071837 - 72081046 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 72920211 - 72929048 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01444130_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NEWGENE_1596188
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008768784.1 XP_008767006.1 524 82
          Gene Symbol: Pigz
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109525.1 NP_001102995.1 3 - 4 446 82
          GenBank mRNA BC161994.1 - 3 - 4 406 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          mRNA splicing, via spliceosome
          7-methylguanosine mRNA capping
          snRNA export from nucleus
          regulation of translation
          regulation of translational initiation
          biological_process
          RNA splicing
          gene silencing by RNA
          mRNA cis splicing, via spliceosome
          positive regulation of RNA export from nucleus
          mRNA transport
          nucleotide binding
          RNA cap binding
          RNA 7-methylguanosine cap binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1019826 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142946 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48355 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1019826 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1019826 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157430 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1019826 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157430 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P150112 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1019826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10754875 - -
          Affymetrix Rat Gene 1.0 ST Array 10754872 - -
          Affymetrix Rat Gene 1.0 ST Array 10754874 - -
          Affymetrix Rat Gene 1.0 ST Array 10754873 - -
          Affymetrix Rat Gene 1.1 ST Array 10754871 - -
          Rat Genome 230 2.0 Array 1385553_at - -
          Rat Genome 230 2.0 Array 1392858_PM_at - -
          Rat Genome 230 2.0 Array 1385553_PM_at - -
          Rat Genome 230 2.0 Array 1392858_at - -
          Rat Genome 230 2.0 Array 1383965_at - -
          Rat Genome 230 2.0 Array 1383965_PM_at - -
          Affymetrix HT Rat Focus Array 1392858_at - -
          Affymetrix HT Rat Focus Array 1385553_PM_at - -
          Affymetrix HT Rat Focus Array 1383965_PM_at - -
          Affymetrix HT Rat Focus Array 1385553_at - -
          Affymetrix HT Rat Focus Array 1392858_PM_at - -
          Affymetrix HT Rat Focus Array 1383965_at - -
          Affymetrix Rat Clariom S Assay PSR1100011194.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011190.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011196.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011188.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011197.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011191.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011189.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011192.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011195.rn.1 TC1100001768.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100011198.rn.1 TC1100001768.rn.1 RTA TC
          Rat Expression Set 230 1383965_at - -
          Rat Expression Set 230 1383965_PM_at - -
          Rat Expression Set 230 1385553_PM_at - -
          Rat Expression Set 230 1392858_at - -
          Rat Expression Set 230 1385553_at - -
          Rat Expression Set 230 1392858_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10754871 - -
          Rat Genome U34 Set rc_AA893307_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21958324 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0