Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01458750_g1
          See other PPIDL1 GE Assays ›
          Gene Symbol:
          Ppidl1 Ppid
          Gene Name:
          peptidylprolyl isomerase D-like 1
          peptidylprolyl isomerase D
          Gene Aliases:
          RGD1560149
          Cyp-40, CypD
          Chromosome Location:
          Chr.9: 121456769 - 121458035 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 178354830 - 178366843 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01458750_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppidl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003750757.3 XP_003750805.1 1 1035 100
          Gene Symbol: Ppid
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004279.1 NP_001004279.1 9 - 10 1087 100
          GenBank mRNA BC076386.1 - 9 - 10 1087 100
          FQ214826.1 - 9 - 10 1090 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001429.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppidl1
          Click here to find siRNAs for Gene Ppid

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          protein folding
          regulation of apoptotic process
          negative regulation of transcription by RNA polymerase II
          apoptotic process
          response to oxidative stress
          protein transport
          lipid droplet organization
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of viral genome replication
          positive regulation of protein secretion
          regulation of mitochondrial membrane potential
          chaperone-mediated protein folding
          protein-containing complex assembly
          cellular response to UV-A
          peptidyl-prolyl cis-trans isomerase activity
          protein binding
          transcription factor binding
          cyclosporin A binding
          enzyme binding
          estrogen receptor binding
          Hsp70 protein binding
          heat shock protein binding
          Hsp90 protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P285225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450147 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69605 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082134 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P124552 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082134 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P124552 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P124552 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082134 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P262229 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P646075 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28054 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816013 - -
          Affymetrix Rat Gene 1.0 ST Array 10816011 - -
          Affymetrix Rat Gene 1.0 ST Array 10816010 - -
          Affymetrix Rat Gene 1.0 ST Array 10816012 - -
          Affymetrix Rat Gene 1.0 ST Array 10926069 - -
          Affymetrix Rat Gene 1.0 ST Array 10816009 - -
          Affymetrix Rat Gene 1.1 ST Array 10926068 - -
          Affymetrix Rat Gene 1.1 ST Array 10816008 - -
          Rat Genome 230 2.0 Array 1380893_PM_at - -
          Rat Genome 230 2.0 Array 1380893_at - -
          Rat Genome 230 2.0 Array 1372827_at - -
          Rat Genome 230 2.0 Array 1380893_at - -
          Rat Genome 230 2.0 Array 1372827_PM_at - -
          Rat Genome 230 2.0 Array 1380893_PM_at - -
          Affymetrix HT Rat Focus Array 1372827_PM_at - -
          Affymetrix HT Rat Focus Array 1372827_at - -
          Affymetrix HT Rat Focus Array 1380893_PM_at - -
          Affymetrix HT Rat Focus Array 1380893_at - -
          Affymetrix Rat Clariom S Assay PSR0200008732.rn.1 TC0200001429.rn.1 RTA TC
          Rat Expression Set 230 1372827_at - -
          Rat Expression Set 230 1372827_PM_at - -
          Rat Expression Set 230 1380893_PM_at - -
          Rat Expression Set 230 1380893_at - -
          Rat Expression Set 230 1380893_PM_at - -
          Rat Expression Set 230 1380893_at - -
          Affymetrix Rat Gene 1.0 ST Array 10926068 - -
          Affymetrix Rat Gene 1.0 ST Array 10816008 - -
          Rat Genome U34 Set rc_AA850781_at - -
          Rat Genome U34 Set rc_AA850781_at - -
          Rat Genome U34 Set rc_AA850781_at - -
          Rat Genome U34 Set rc_AA850781_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22179938 - -
          ABI Rat Genome Survey Microarray 22368762 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354575 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361286 - -
          RatRef-12 Expression BeadChip ILMN_55665 - -
          RatRef-12 Expression BeadChip ILMN_55687 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0