Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460084_g1
          See other PPP2CA GE Assays ›
          Gene Symbol:
          Ppp2ca LOC103694903
          Gene Name:
          protein phosphatase 2, catalytic subunit, alpha isozyme
          serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform
          Gene Aliases:
          Pp2a1
          -
          Chromosome Location:
          Chr.10: 37535108 - 37554861 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906910: 112829 - 133730 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01460084_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp2ca
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017039.2 NP_058735.1 6 - 7 1055 84
          GenBank mRNA BC070914.1 - 6 - 7 1064 84
          BC072531.1 - 6 - 7 1055 84
          FQ228785.1 - 5 - 6 619 84
          M33114.1 - 6 - 7 971 84
          X14159.1 - 6 - 7 971 84
          X16043.1 - 6 - 7 1030 84
          Gene Symbol: LOC103694903
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008773894.1 XP_008772116.1 1550 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp2ca
          Click here to find siRNAs for Gene LOC103694903

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          regulation of protein phosphorylation
          negative regulation of protein phosphorylation
          cardiac ventricle development
          protein dephosphorylation
          mesoderm development
          heart development
          regulation of signaling receptor activity
          negative regulation of epithelial to mesenchymal transition
          regulation of protein autophosphorylation
          positive regulation of phosphoprotein phosphatase activity
          cellular response to insulin stimulus
          positive regulation of protein dephosphorylation
          regulation of protein catabolic process
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          meiotic cell cycle
          protein heterotrimerization
          cellular response to calcium ion
          cellular response to glucose stimulus
          cellular response to cytokine stimulus
          cellular response to ethanol
          cellular response to follicle-stimulating hormone stimulus
          cellular response to steroid hormone stimulus
          positive regulation of protein serine/threonine kinase activity
          negative regulation of calcium ion transmembrane transporter activity
          protein serine/threonine phosphatase activity
          protein binding
          protein C-terminus binding
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          protein domain specific binding
          protein kinase B binding
          transmembrane transporter binding
          protein-containing complex binding
          metal ion binding
          protein heterodimerization activity
          protein dimerization activity
          tau protein binding
          GABA receptor binding
          protein phosphatase 2A binding
          protein antigen binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P273989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178355 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P273989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P273989 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P273989 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P273990 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P273989 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701192964 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61442 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50741 - -
          Agilent Rat cDNA Microarray (G4105A) 701192964 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10752269 - -
          Affymetrix Rat Gene 1.0 ST Array 10733424 - -
          Affymetrix Rat Gene 1.0 ST Array 10733423 - -
          Affymetrix Rat Gene 1.0 ST Array 10733420 - -
          Affymetrix Rat Gene 1.0 ST Array 10733419 - -
          Affymetrix Rat Gene 1.0 ST Array 10733426 - -
          Affymetrix Rat Gene 1.0 ST Array 10733422 - -
          Affymetrix Rat Gene 1.0 ST Array 10733421 - -
          Affymetrix Rat Gene 1.0 ST Array 10733425 - -
          Affymetrix Rat Gene 1.0 ST Array 10733418 - -
          Affymetrix Rat Gene 1.1 ST Array 10733417 - -
          Affymetrix Rat Gene 1.1 ST Array 10752268 - -
          Rat Genome 230 2.0 Array 1398790_PM_at - -
          Rat Genome 230 2.0 Array 1380061_at - -
          Rat Genome 230 2.0 Array 1380061_PM_at - -
          Rat Genome 230 2.0 Array 1388805_PM_at - -
          Rat Genome 230 2.0 Array 1388805_at - -
          Rat Genome 230 2.0 Array 1398790_at - -
          Affymetrix HT Rat Focus Array 1380061_PM_at - -
          Affymetrix HT Rat Focus Array 1380061_at - -
          Affymetrix HT Rat Focus Array 1398790_at - -
          Affymetrix HT Rat Focus Array 1398790_PM_at - -
          Rat Expression Set 230 1398790_PM_at - -
          Rat Expression Set 230 1398790_at - -
          Rat Expression Set 230 1388805_PM_at - -
          Rat Expression Set 230 1388805_at - -
          Rat Expression Set 230 1380061_PM_at - -
          Rat Expression Set 230 1380061_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733417 - -
          Affymetrix Rat Gene 1.0 ST Array 10752268 - -
          Rat Genome U34 Set rc_AI012595_at - -
          Rat Genome U34 Set X16043cds_at - -
          Rat Genome U34 Set rc_AI059014_f_at - -
          Rat Genome U34 Set rc_AI009467_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21704895 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358097 - -
          RatRef-12 Expression BeadChip ILMN_64026 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0