Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460878_m1
          See other DHX58 GE Assays ›
          Gene Symbol:
          Dhx58
          Gene Name:
          DEXH (Asp-Glu-X-His) box polypeptide 58
          Gene Aliases:
          Lgp2, RGD1310093
          Chromosome Location:
          Chr.10: 88599722 - 88611565 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460878_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dhx58
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001098788.1 NP_001092258.1 10 - 11 1752 62
          XM_006247314.2 XP_006247376.1 11 - 12 2321 62
          XM_006247315.2 XP_006247377.1 11 - 12 2310 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003805.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          response to virus
          negative regulation of type I interferon production
          positive regulation of type I interferon production
          negative regulation of MDA-5 signaling pathway
          negative regulation of RIG-I signaling pathway
          regulation of innate immune response
          negative regulation of innate immune response
          positive regulation of MDA-5 signaling pathway
          positive regulation of RIG-I signaling pathway
          DNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          ATP binding
          zinc ion binding
          hydrolase activity, acting on acid anhydrides

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P165403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57968 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P165403 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P165403 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21799 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20957 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10747431 - -
          Affymetrix Rat Gene 1.0 ST Array 10747427 - -
          Affymetrix Rat Gene 1.0 ST Array 10747433 - -
          Affymetrix Rat Gene 1.0 ST Array 10747435 - -
          Affymetrix Rat Gene 1.0 ST Array 10747430 - -
          Affymetrix Rat Gene 1.0 ST Array 10747434 - -
          Affymetrix Rat Gene 1.0 ST Array 10747438 - -
          Affymetrix Rat Gene 1.0 ST Array 10747429 - -
          Affymetrix Rat Gene 1.0 ST Array 10747436 - -
          Affymetrix Rat Gene 1.0 ST Array 10747432 - -
          Affymetrix Rat Gene 1.0 ST Array 10747437 - -
          Affymetrix Rat Gene 1.0 ST Array 10747428 - -
          Affymetrix Rat Gene 1.1 ST Array 10747426 - -
          Rat Genome 230 2.0 Array 1385277_PM_x_at - -
          Rat Genome 230 2.0 Array 1385277_x_at - -
          Rat Genome 230 2.0 Array 1385439_PM_x_at - -
          Rat Genome 230 2.0 Array 1385439_x_at - -
          Rat Genome 230 2.0 Array 1385276_PM_a_at - -
          Rat Genome 230 2.0 Array 1385276_a_at - -
          Affymetrix HT Rat Focus Array 1385276_PM_a_at - -
          Affymetrix HT Rat Focus Array 1385277_x_at - -
          Affymetrix HT Rat Focus Array 1385277_PM_x_at - -
          Affymetrix HT Rat Focus Array 1385276_a_at - -
          Affymetrix Rat Clariom S Assay PSR1000034115.rn.1 TC1000003805.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034114.rn.1 TC1000003805.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000034113.rn.1 TC1000003805.rn.1 RTA TC
          Rat Expression Set 230 1385276_a_at - -
          Rat Expression Set 230 1385277_PM_x_at - -
          Rat Expression Set 230 1385439_x_at - -
          Rat Expression Set 230 1385277_x_at - -
          Rat Expression Set 230 1385439_PM_x_at - -
          Rat Expression Set 230 1385276_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10747426 - -
          Rat Genome U34 Set rc_AA818849_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21892413 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372498 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0