Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01462610_m1
          See other PLCZ1 GE Assays ›
          Gene Symbol:
          Plcz1
          Gene Name:
          phospholipase C, zeta 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 174125072 - 174181525 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01462610_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plcz1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012234.1 NP_001012234.1 11 - 12 1703 98
          XM_006237597.2 XP_006237659.1 12 - 13 1932 98
          XM_006237600.2 XP_006237662.1 11 - 12 1808 98
          GenBank mRNA AY885259.1 - 11 - 12 1703 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400004004.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plcz1

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          multicellular organism development
          egg activation
          lipid catabolic process
          intracellular signal transduction
          phosphatidylinositol phospholipase C activity
          phospholipase C activity
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P791053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108968 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147894 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132392 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48122 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154105 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P791053 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P791053 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P791053 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P791053 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P959571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78321 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11410 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103735 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866590 - -
          Affymetrix Rat Gene 1.0 ST Array 10866593 - -
          Affymetrix Rat Gene 1.0 ST Array 10866601 - -
          Affymetrix Rat Gene 1.0 ST Array 10866591 - -
          Affymetrix Rat Gene 1.0 ST Array 10866599 - -
          Affymetrix Rat Gene 1.0 ST Array 10866597 - -
          Affymetrix Rat Gene 1.0 ST Array 10866589 - -
          Affymetrix Rat Gene 1.0 ST Array 10866595 - -
          Affymetrix Rat Gene 1.0 ST Array 10866602 - -
          Affymetrix Rat Gene 1.0 ST Array 10866600 - -
          Affymetrix Rat Gene 1.0 ST Array 10866594 - -
          Affymetrix Rat Gene 1.0 ST Array 10866598 - -
          Affymetrix Rat Gene 1.0 ST Array 10866592 - -
          Affymetrix Rat Gene 1.0 ST Array 10866596 - -
          Affymetrix Rat Gene 1.1 ST Array 10866588 - -
          Affymetrix Rat Clariom S Assay PSR0400028372.rn.1 TC0400004004.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028373.rn.1 TC0400004004.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10866588 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21705312 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0