Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01465899_m1
          See other SUCLA2 GE Assays ›
          Gene Symbol:
          Sucla2
          Gene Name:
          succinate-CoA ligase, ADP-forming, beta subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 55461695 - 55516954 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01465899_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sucla2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108387.2 NP_001101857.2 4 - 5 540 81
          GenBank mRNA BC166998.1 - 3 - 4 460 81
          BF408124.1 - 4 - 5 504 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000824.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          tricarboxylic acid cycle
          succinyl-CoA metabolic process
          succinate metabolic process
          succinate-CoA ligase (ADP-forming) activity
          ATP binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P417104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100658 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83674 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142197 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112469 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143651 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P417104 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P417104 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P417104 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P108982 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P417104 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32104 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97063 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781459 - -
          Affymetrix Rat Gene 1.0 ST Array 10781454 - -
          Affymetrix Rat Gene 1.0 ST Array 10781456 - -
          Affymetrix Rat Gene 1.0 ST Array 10781455 - -
          Affymetrix Rat Gene 1.0 ST Array 10781452 - -
          Affymetrix Rat Gene 1.0 ST Array 10781462 - -
          Affymetrix Rat Gene 1.0 ST Array 10781457 - -
          Affymetrix Rat Gene 1.0 ST Array 10781460 - -
          Affymetrix Rat Gene 1.0 ST Array 10781458 - -
          Affymetrix Rat Gene 1.0 ST Array 10781461 - -
          Affymetrix Rat Gene 1.0 ST Array 10781453 - -
          Affymetrix Rat Gene 1.1 ST Array 10781451 - -
          Rat Genome 230 2.0 Array 1388975_at - -
          Rat Genome 230 2.0 Array 1388975_PM_at - -
          Rat Genome 230 2.0 Array 1383438_at - -
          Rat Genome 230 2.0 Array 1383438_PM_at - -
          Rat Genome 230 2.0 Array 1381671_at - -
          Rat Genome 230 2.0 Array 1381671_PM_at - -
          Affymetrix HT Rat Focus Array 1383438_at - -
          Affymetrix HT Rat Focus Array 1381671_PM_at - -
          Affymetrix HT Rat Focus Array 1388975_at - -
          Affymetrix HT Rat Focus Array 1388975_PM_at - -
          Affymetrix HT Rat Focus Array 1381671_at - -
          Affymetrix HT Rat Focus Array 1383438_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500005665.rn.1 TC1500000824.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005667.rn.1 TC1500000824.rn.1 RTA TC
          Rat Expression Set 230 1383438_at - -
          Rat Expression Set 230 1388975_at - -
          Rat Expression Set 230 1388975_PM_at - -
          Rat Expression Set 230 1383438_PM_at - -
          Rat Expression Set 230 1381671_PM_at - -
          Rat Expression Set 230 1381671_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781451 - -
          Rat Genome U34 Set rc_AA800194_at - -
          Rat Genome U34 Set rc_AA923982_g_at - -
          Rat Genome U34 Set rc_AA923982_at - -
          Rat Genome U34 Set rc_AI169320_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22166992 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357638 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-77bd865d8c-25mtt:80/100.66.130.62:80.
          git-commit: 6e13e51bb2b364e4bd246c4dbbae92ef278be7e6
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline