Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01468484_m1
          See other ATL2 GE Assays ›
          Gene Symbol:
          Atl2
          Gene Name:
          atlastin GTPase 2
          Gene Aliases:
          Arl6ip2
          Chromosome Location:
          Chr.6: 2568917 - 2610432 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01468484_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100671.1 NP_001094141.1 11 - 12 1196 67
          XM_006239640.2 XP_006239702.1 11 - 12 1234 67
          XM_006239641.2 XP_006239703.1 11 - 12 1294 67
          GenBank mRNA EV762626.1 - 2 - 3 145 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000031.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          endoplasmic reticulum to Golgi vesicle-mediated transport
          endoplasmic reticulum organization
          Golgi organization
          metabolic process
          protein homooligomerization
          GTPase activity
          GTP binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P701991 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P086259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69692 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086259 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P701991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P701991 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P086259 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P701991 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P213346 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10887964 - -
          Affymetrix Rat Gene 1.0 ST Array 10887957 - -
          Affymetrix Rat Gene 1.0 ST Array 10887955 - -
          Affymetrix Rat Gene 1.0 ST Array 10887956 - -
          Affymetrix Rat Gene 1.0 ST Array 10887960 - -
          Affymetrix Rat Gene 1.0 ST Array 10887963 - -
          Affymetrix Rat Gene 1.0 ST Array 10887959 - -
          Affymetrix Rat Gene 1.0 ST Array 10887958 - -
          Affymetrix Rat Gene 1.0 ST Array 10887965 - -
          Affymetrix Rat Gene 1.0 ST Array 10887962 - -
          Affymetrix Rat Gene 1.0 ST Array 10887954 - -
          Affymetrix Rat Gene 1.0 ST Array 10887961 - -
          Affymetrix Rat Gene 1.1 ST Array 10887952 - -
          Rat Genome 230 2.0 Array 1384131_at - -
          Rat Genome 230 2.0 Array 1372085_at - -
          Rat Genome 230 2.0 Array 1384131_PM_at - -
          Rat Genome 230 2.0 Array 1372085_PM_at - -
          Affymetrix HT Rat Focus Array 1372085_PM_at - -
          Affymetrix HT Rat Focus Array 1372085_at - -
          Affymetrix HT Rat Focus Array 1384131_at - -
          Affymetrix HT Rat Focus Array 1384131_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600000242.rn.1 TC0600000031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000235.rn.1 TC0600000031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000241.rn.1 TC0600000031.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000243.rn.1 TC0600000031.rn.1 RTA TC
          Rat Expression Set 230 1372085_at - -
          Rat Expression Set 230 1372085_PM_at - -
          Rat Expression Set 230 1384131_at - -
          Rat Expression Set 230 1384131_PM_at - -
          Rat Genome U34 Set rc_AI171587_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22132709 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363949 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0