Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474494_g1
          See other DOC2G GE Assays ›
          Gene Symbol:
          Doc2g
          Gene Name:
          double C2-like domains, gamma
          Gene Aliases:
          Doc2gp
          Chromosome Location:
          Chr.1: 219249247 - 219253790 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01474494_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Doc2g
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011937.1 NP_001011937.1 10 - 11 1167 84
          XM_006230735.2 XP_006230797.1 10 - 11 2191 84
          GenBank mRNA BC083779.1 - 10 - 11 1167 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002991.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Doc2g

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P447810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225838 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P447810 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P447810 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P447810 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P447810 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10712775 - -
          Affymetrix Rat Gene 1.0 ST Array 10712767 - -
          Affymetrix Rat Gene 1.0 ST Array 10712768 - -
          Affymetrix Rat Gene 1.0 ST Array 10712773 - -
          Affymetrix Rat Gene 1.0 ST Array 10712771 - -
          Affymetrix Rat Gene 1.0 ST Array 10712774 - -
          Affymetrix Rat Gene 1.0 ST Array 10712770 - -
          Affymetrix Rat Gene 1.0 ST Array 10712772 - -
          Affymetrix Rat Gene 1.0 ST Array 10712765 - -
          Affymetrix Rat Gene 1.0 ST Array 10712769 - -
          Affymetrix Rat Gene 1.0 ST Array 10712766 - -
          Affymetrix Rat Gene 1.1 ST Array 10712763 - -
          Rat Genome 230 2.0 Array 1382327_at - -
          Rat Genome 230 2.0 Array 1382327_PM_at - -
          Affymetrix HT Rat Focus Array 1382327_at - -
          Affymetrix HT Rat Focus Array 1382327_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100024105.rn.1 TC0100002991.rn.1 RTA TC
          Rat Expression Set 230 1382327_at - -
          Rat Expression Set 230 1382327_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10712763 - -
          Rat Genome U34 Set rc_AI104824_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21332124 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376478 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0