Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01479211_m1
          See other CYB561A3 GE Assays ›
          Gene Symbol:
          Cyb561a3
          Gene Name:
          cytochrome b561 family, member A3
          Gene Aliases:
          Cybasc3, RGD1304670
          Chromosome Location:
          Chr.1: 226620933 - 226642246 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01479211_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyb561a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014164.1 NP_001014186.1 4 - 5 604 57
          XM_006231049.2 XP_006231111.1 4 - 5 1204 57
          XM_006231050.2 XP_006231112.1 4 - 5 1072 57
          XM_006231051.2 XP_006231113.1 4 - 5 1037 57
          XM_006231052.2 XP_006231114.1 4 - 5 1239 57
          XM_006231053.2 XP_006231115.1 4 - 5 623 57
          XM_006231054.2 XP_006231116.1 4 - 5 583 57
          XM_006231055.2 XP_006231117.1 3 - 4 786 57
          XM_008760233.1 XP_008758455.1 4 - 5 959 57
          XM_008760234.1 XP_008758456.1 4 - 5 567 57
          GenBank mRNA BC085835.1 - 4 - 5 604 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003202.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyb561a3

          Gene Ontology Categories:

          Process(es)

          oxidoreductase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P557680 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P896040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97639 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64523 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P896040 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557680 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P557680 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P896040 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P557680 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P896040 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557680 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P896040 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37380 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 148 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8014 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713913 - -
          Affymetrix Rat Gene 1.0 ST Array 10713912 - -
          Affymetrix Rat Gene 1.0 ST Array 10713911 - -
          Affymetrix Rat Gene 1.0 ST Array 10713908 - -
          Affymetrix Rat Gene 1.0 ST Array 10713909 - -
          Affymetrix Rat Gene 1.0 ST Array 10713914 - -
          Affymetrix Rat Gene 1.0 ST Array 10713910 - -
          Affymetrix Rat Gene 1.0 ST Array 10713906 - -
          Affymetrix Rat Gene 1.0 ST Array 10713905 - -
          Affymetrix Rat Gene 1.0 ST Array 10713907 - -
          Affymetrix Rat Gene 1.1 ST Array 10713904 - -
          Rat Genome 230 2.0 Array 1372846_PM_at - -
          Rat Genome 230 2.0 Array 1372846_at - -
          Affymetrix HT Rat Focus Array 1372846_at - -
          Affymetrix HT Rat Focus Array 1372846_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100026447.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026457.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026463.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026459.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026462.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026437.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026453.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026454.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026455.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026460.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026461.rn.1 TC0100003202.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100026458.rn.1 TC0100003202.rn.1 RTA TC
          Rat Expression Set 230 1372846_PM_at - -
          Rat Expression Set 230 1372846_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713904 - -
          Rat Genome U34 Set rc_AI179227_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22384292 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650159 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0