Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481368_m1
          See other FAM101A GE Assays ›
          Gene Symbol:
          Fam101a
          Gene Name:
          family with sequence similarity 101, member A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 37038732 - 37047628 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481368_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam101a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109547.1 NP_001103017.1 2 - 3 411 63
          XM_008769261.1 XP_008767483.1 2 - 3 161 63
          XM_008769262.1 XP_008767484.1 2 - 3 747 63
          GenBank mRNA FQ235329.1 - 2 - 3 200 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001524.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system morphogenesis
          negative regulation of chondrocyte development
          actin filament bundle organization
          negative regulation of bone mineralization involved in bone maturation
          filamin binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P077820 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P638638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82404 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20609 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P638638 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P077820 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P638638 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P077820 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P077820 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P638638 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10761552 - -
          Affymetrix Rat Gene 1.0 ST Array 10761550 - -
          Affymetrix Rat Gene 1.0 ST Array 10761551 - -
          Affymetrix Rat Gene 1.1 ST Array 10761549 - -
          Rat Genome 230 2.0 Array 1385555_PM_at - -
          Rat Genome 230 2.0 Array 1385555_at - -
          Affymetrix Rat Clariom S Assay PSR1200012670.rn.1 TC1200001524.rn.1 RTA TC
          Rat Expression Set 230 1385555_at - -
          Rat Expression Set 230 1385555_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10761549 - -
          Rat Genome U34 Set rc_AA924444_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20803940 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0