Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01483937_m1
          See other POGK GE Assays ›
          Gene Symbol:
          Pogk
          Gene Name:
          pogo transposable element with KRAB domain
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 84603126 - 84619083 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01483937_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pogk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107194.2 NP_001100664.2 4 - 5 457 154
          XM_006250161.2 XP_006250223.1 5 - 6 831 154
          XM_006250162.2 XP_006250224.1 5 - 6 696 154

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001847.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P088910 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P088905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50087 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175561 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78179 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088905 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088910 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088910 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088905 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P088905 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P088910 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13566 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769598 - -
          Affymetrix Rat Gene 1.0 ST Array 10769599 - -
          Affymetrix Rat Gene 1.0 ST Array 10769596 - -
          Affymetrix Rat Gene 1.0 ST Array 10769600 - -
          Affymetrix Rat Gene 1.0 ST Array 10769597 - -
          Affymetrix Rat Gene 1.1 ST Array 10769595 - -
          Rat Genome 230 2.0 Array 1379731_at - -
          Rat Genome 230 2.0 Array 1385313_at - -
          Rat Genome 230 2.0 Array 1385313_PM_at - -
          Rat Genome 230 2.0 Array 1379731_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300013060.rn.1 TC1300001847.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013057.rn.1 TC1300001847.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013061.rn.1 TC1300001847.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013062.rn.1 TC1300001847.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013058.rn.1 TC1300001847.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300013059.rn.1 TC1300001847.rn.1 RTA TC
          Rat Expression Set 230 1385313_at - -
          Rat Expression Set 230 1385313_PM_at - -
          Rat Expression Set 230 1379731_PM_at - -
          Rat Expression Set 230 1379731_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769595 - -
          Rat Genome U34 Set rc_AA851877_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20874430 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356636 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0