Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01488426_m1
          See other CNOT7 GE Assays ›
          Gene Symbol:
          Cnot7
          Gene Name:
          CCR4-NOT transcription complex, subunit 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 54899452 - 54918616 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01488426_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnot7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107313.1 NP_001100783.1 6 - 7 1022 78
          XM_008771290.1 XP_008769512.1 6 - 7 1195 78
          XM_008771291.1 XP_008769513.1 6 - 7 1165 78
          XM_008771292.1 XP_008769514.1 6 - 7 1065 78
          GenBank mRNA BC167766.1 - 6 - 7 981 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000634.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          nuclear-transcribed mRNA poly(A) tail shortening
          deadenylation-dependent decapping of nuclear-transcribed mRNA
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          negative regulation of gene expression
          gene silencing by RNA
          P-body assembly
          exonucleolytic catabolism of deadenylated mRNA
          positive regulation of transcription by RNA polymerase II
          positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
          positive regulation of mRNA catabolic process
          RNA phosphodiester bond hydrolysis, exonucleolytic
          positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          3'-5'-exoribonuclease activity
          nucleic acid binding
          exoribonuclease activity
          poly(A)-specific ribonuclease activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1020018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16649 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1020018 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1020018 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1020018 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1020018 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P489839 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80598 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10788419 - -
          Affymetrix Rat Gene 1.0 ST Array 10788416 - -
          Affymetrix Rat Gene 1.0 ST Array 10788417 - -
          Affymetrix Rat Gene 1.0 ST Array 10787211 - -
          Affymetrix Rat Gene 1.0 ST Array 10788415 - -
          Affymetrix Rat Gene 1.0 ST Array 10788420 - -
          Affymetrix Rat Gene 1.0 ST Array 10788418 - -
          Affymetrix Rat Gene 1.1 ST Array 10787209 - -
          Affymetrix Rat Gene 1.1 ST Array 10788414 - -
          Rat Genome 230 2.0 Array 1367515_PM_at - -
          Rat Genome 230 2.0 Array 1367515_at - -
          Affymetrix HT Rat Focus Array 1367515_PM_at - -
          Affymetrix HT Rat Focus Array 1367515_at - -
          Affymetrix Rat Clariom S Assay PSR1600005064.rn.1 TC1600000634.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600005063.rn.1 TC1600000634.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600005065.rn.1 TC1600000634.rn.1 RTA TC
          Rat Expression Set 230 1367515_at - -
          Rat Expression Set 230 1367515_PM_at - -
          Rat Expression Set 230 1367515_at - -
          Rat Expression Set 230 1367515_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10788414 - -
          Rat Genome U34 Set rc_AI071980_at - -
          Rat Genome U34 Set rc_AI101934_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20824790 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353588 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0