Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc21a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF445994.1 | - | 2 - 3 | 146 | 133 | |
AF445995.1 | - | 2 - 3 | 146 | 133 | ||
AF445996.1 | - | 2 - 3 | 146 | 133 | ||
AF445997.1 | - | 2 - 3 | 146 | 133 |
Set Membership: |
![]() ![]() ![]() |