Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01494304_g1
          See other NEWGENE_1310139 GE Assays ›
          Gene Symbol:
          NEWGENE_1310139 Mast4
          Gene Name:
          microtubule associated serine/threonine kinase family member 4
          microtubule associated serine/threonine kinase family member 4
          Gene Aliases:
          Mast4
          RGD1310139
          Chromosome Location:
          Chr.2: 32857576 - 33026430 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 32442285 - 32519052 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01494304_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NEWGENE_1310139
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008760704.1 XP_008758926.1 25 - 26 4742 104
          XM_008760705.1 XP_008758927.1 24 - 25 4541 104
          Gene Symbol: Mast4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008760713.1 XP_008758935.1 3394 104

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          magnesium ion binding
          protein serine/threonine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17397 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1033985 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17397 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17397 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17397 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1033985 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17397 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P623197 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821102 - -
          Affymetrix Rat Gene 1.0 ST Array 10821112 - -
          Affymetrix Rat Gene 1.0 ST Array 10821111 - -
          Affymetrix Rat Gene 1.0 ST Array 10821100 - -
          Affymetrix Rat Gene 1.0 ST Array 10821106 - -
          Affymetrix Rat Gene 1.0 ST Array 10821101 - -
          Affymetrix Rat Gene 1.0 ST Array 10821099 - -
          Affymetrix Rat Gene 1.0 ST Array 10821110 - -
          Affymetrix Rat Gene 1.0 ST Array 10821103 - -
          Affymetrix Rat Gene 1.0 ST Array 10821109 - -
          Affymetrix Rat Gene 1.0 ST Array 10821104 - -
          Rat Genome 230 2.0 Array 1390658_PM_at - -
          Rat Genome 230 2.0 Array 1377972_PM_at - -
          Rat Genome 230 2.0 Array 1390658_at - -
          Rat Genome 230 2.0 Array 1375313_PM_at - -
          Rat Genome 230 2.0 Array 1391777_PM_at - -
          Rat Genome 230 2.0 Array 1390905_at - -
          Rat Genome 230 2.0 Array 1377972_at - -
          Rat Genome 230 2.0 Array 1378969_at - -
          Rat Genome 230 2.0 Array 1391777_at - -
          Rat Genome 230 2.0 Array 1378969_PM_at - -
          Rat Genome 230 2.0 Array 1375313_at - -
          Rat Genome 230 2.0 Array 1390905_PM_at - -
          Affymetrix HT Rat Focus Array 1375313_PM_at - -
          Affymetrix HT Rat Focus Array 1390658_PM_at - -
          Affymetrix HT Rat Focus Array 1377972_at - -
          Affymetrix HT Rat Focus Array 1390658_at - -
          Affymetrix HT Rat Focus Array 1377972_PM_at - -
          Affymetrix HT Rat Focus Array 1375313_at - -
          Rat Expression Set 230 1390658_at - -
          Rat Expression Set 230 1375313_at - -
          Rat Expression Set 230 1375313_PM_at - -
          Rat Expression Set 230 1390658_PM_at - -
          Rat Expression Set 230 1378969_PM_at - -
          Rat Expression Set 230 1390905_at - -
          Rat Expression Set 230 1378969_at - -
          Rat Expression Set 230 1377972_PM_at - -
          Rat Expression Set 230 1390905_PM_at - -
          Rat Expression Set 230 1377972_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821105 - -
          Rat Genome U34 Set rc_AI044530_at - -
          Rat Genome U34 Set rc_AI177282_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21399438 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357383 - -
          RatRef-12 Expression BeadChip ILMN_64077 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix