Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497309_g1
          See other PBDC1 GE Assays ›
          Gene Symbol:
          Pbdc1
          Gene Name:
          polysaccharide biosynthesis domain containing 1
          Gene Aliases:
          RGD1562502
          Chromosome Location:
          Chr.X: 76042239 - 76092964 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497309_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pbdc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108818.1 NP_001102288.1 5 - 6 1066 81
          GenBank mRNA BC166918.1 - 5 - 6 429 81
          FQ220252.1 - 5 - 6 430 81
          FQ225636.1 - 5 - 6 449 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000701.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1015178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73888 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1015178 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1015178 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1015178 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1015178 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934551 - -
          Affymetrix Rat Gene 1.0 ST Array 10934554 - -
          Affymetrix Rat Gene 1.0 ST Array 10934553 - -
          Affymetrix Rat Gene 1.0 ST Array 10934549 - -
          Affymetrix Rat Gene 1.0 ST Array 10934550 - -
          Affymetrix Rat Gene 1.0 ST Array 10934552 - -
          Affymetrix Rat Gene 1.1 ST Array 10934548 - -
          Rat Genome 230 2.0 Array 1372543_PM_at - -
          Rat Genome 230 2.0 Array 1372543_at - -
          Affymetrix HT Rat Focus Array 1372543_at - -
          Affymetrix HT Rat Focus Array 1372543_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00004462.rn.1 TC0X00000701.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00004460.rn.1 TC0X00000701.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00004461.rn.1 TC0X00000701.rn.1 RTA TC
          Rat Expression Set 230 1372543_PM_at - -
          Rat Expression Set 230 1372543_at - -
          Affymetrix Rat Gene 1.0 ST Array 10934548 - -
          Rat Genome U34 Set rc_AA965113_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21626337 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358626 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix