Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01497777_m1
          See other AP3B1 GE Assays ›
          Gene Symbol:
          Ap3b1
          Gene Name:
          adaptor-related protein complex 3, beta 1 subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 24022896 - 24227521 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01497777_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ap3b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107646.1 NP_001101116.1 22 - 23 2720 65
          XM_006231814.2 XP_006231876.1 24 - 25 4909 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000178.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to lysosome
          blood coagulation
          anterograde axonal transport
          biological_process
          antigen processing and presentation
          melanosome organization
          antigen processing and presentation, exogenous lipid antigen via MHC class Ib
          anterograde synaptic vesicle transport
          positive regulation of NK T cell differentiation
          molecular_function
          protein phosphatase binding
          GTP-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P701756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72212 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89018 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96458 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4178 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66257 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39284 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88283 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76125 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99246 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158582 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118442 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175581 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3413 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P701756 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P701756 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P701756 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P189785 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40124 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85279 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66164 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40230 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10812538 - -
          Affymetrix Rat Gene 1.0 ST Array 10812533 - -
          Affymetrix Rat Gene 1.0 ST Array 10812555 - -
          Affymetrix Rat Gene 1.0 ST Array 10812542 - -
          Affymetrix Rat Gene 1.0 ST Array 10812559 - -
          Affymetrix Rat Gene 1.0 ST Array 10812547 - -
          Affymetrix Rat Gene 1.0 ST Array 10812544 - -
          Affymetrix Rat Gene 1.0 ST Array 10812554 - -
          Affymetrix Rat Gene 1.0 ST Array 10812558 - -
          Affymetrix Rat Gene 1.0 ST Array 10812550 - -
          Affymetrix Rat Gene 1.0 ST Array 10812543 - -
          Affymetrix Rat Gene 1.0 ST Array 10812556 - -
          Affymetrix Rat Gene 1.0 ST Array 10812536 - -
          Affymetrix Rat Gene 1.0 ST Array 10812553 - -
          Affymetrix Rat Gene 1.0 ST Array 10812549 - -
          Affymetrix Rat Gene 1.0 ST Array 10812546 - -
          Affymetrix Rat Gene 1.0 ST Array 10812552 - -
          Affymetrix Rat Gene 1.0 ST Array 10812557 - -
          Affymetrix Rat Gene 1.0 ST Array 10812537 - -
          Affymetrix Rat Gene 1.0 ST Array 10812545 - -
          Affymetrix Rat Gene 1.0 ST Array 10812539 - -
          Affymetrix Rat Gene 1.0 ST Array 10812548 - -
          Affymetrix Rat Gene 1.0 ST Array 10812551 - -
          Affymetrix Rat Gene 1.0 ST Array 10812541 - -
          Affymetrix Rat Gene 1.1 ST Array 10812532 - -
          Rat Genome 230 2.0 Array 1373888_at - -
          Rat Genome 230 2.0 Array 1373888_PM_at - -
          Affymetrix HT Rat Focus Array 1373888_at - -
          Affymetrix HT Rat Focus Array 1373888_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200000946.rn.1 TC0200000178.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200000947.rn.1 TC0200000178.rn.1 RTA TC
          Rat Expression Set 230 1373888_PM_at - -
          Rat Expression Set 230 1373888_at - -
          Affymetrix Rat Gene 1.0 ST Array 10812532 - -
          Rat Genome U34 Set rc_AI170332_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20810747 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359820 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0