Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01498359_gH
          See other RAB1B GE Assays ›
          Gene Symbol:
          Rab1b LOC102555167
          Gene Name:
          RAB1B, member RAS oncogene family
          ras-related protein Rab-1B-like
          Gene Aliases:
          Rab1bl
          -
          Chromosome Location:
          Chr.1: 220483529 - 220491469 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 220480828 - 220482648 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01498359_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109979.2 NP_001103449.1 486 94
          GenBank mRNA BC085118.1 - 453 94
          CB714630.1 - 131 94
          DN934711.1 - 504 94
          EV762710.1 - 505 94
          X13905.1 - 426 94
          Gene Symbol: LOC102555167
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008760197.1 XP_008758419.1 1 454 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007103.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          endoplasmic reticulum to Golgi vesicle-mediated transport
          metabolic process
          virion assembly
          Rab protein signal transduction
          positive regulation of glycoprotein metabolic process
          regulation of autophagosome assembly
          GTPase activity
          GTP binding
          GDP binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069769 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P002909 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P051802 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P069764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369044 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153461 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002909 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069769 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069764 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051802 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069769 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P069764 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P051802 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002909 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P002909 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P051802 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069764 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P069769 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10727738 - -
          Affymetrix Rat Gene 1.0 ST Array 10727736 - -
          Affymetrix Rat Gene 1.0 ST Array 10727739 - -
          Affymetrix Rat Gene 1.0 ST Array 10727737 - -
          Affymetrix Rat Gene 1.0 ST Array 10727735 - -
          Affymetrix Rat Gene 1.0 ST Array 10933946 - -
          Affymetrix Rat Gene 1.1 ST Array 10933945 - -
          Affymetrix Rat Gene 1.1 ST Array 10727734 - -
          Rat Genome 230 2.0 Array 1398982_PM_at - -
          Rat Genome 230 2.0 Array 1385270_s_at - -
          Rat Genome 230 2.0 Array 1385206_PM_at - -
          Rat Genome 230 2.0 Array 1392855_x_at - -
          Rat Genome 230 2.0 Array 1382946_PM_a_at - -
          Rat Genome 230 2.0 Array 1385206_at - -
          Rat Genome 230 2.0 Array 1382947_PM_at - -
          Rat Genome 230 2.0 Array 1393669_at - -
          Rat Genome 230 2.0 Array 1382947_at - -
          Rat Genome 230 2.0 Array 1385270_PM_s_at - -
          Rat Genome 230 2.0 Array 1393669_PM_at - -
          Rat Genome 230 2.0 Array 1382946_a_at - -
          Rat Genome 230 2.0 Array 1392855_PM_x_at - -
          Rat Genome 230 2.0 Array 1398982_at - -
          Affymetrix HT Rat Focus Array 1392855_x_at - -
          Affymetrix HT Rat Focus Array 1392855_PM_x_at - -
          Affymetrix HT Rat Focus Array 1385206_PM_at - -
          Affymetrix HT Rat Focus Array 1398982_PM_at - -
          Affymetrix HT Rat Focus Array 1385206_at - -
          Affymetrix HT Rat Focus Array 1382946_PM_a_at - -
          Affymetrix HT Rat Focus Array 1398982_at - -
          Affymetrix HT Rat Focus Array 1382946_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100056659.rn.1 TC0100007103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100056658.rn.1 TC0100007103.rn.1 RTA TC
          Rat Expression Set 230 1398982_at - -
          Rat Expression Set 230 1393669_PM_at - -
          Rat Expression Set 230 1385206_at - -
          Rat Expression Set 230 1393669_at - -
          Rat Expression Set 230 1385206_PM_at - -
          Rat Expression Set 230 1398982_PM_at - -
          Rat Expression Set 230 1382947_PM_at - -
          Rat Expression Set 230 1385270_PM_s_at - -
          Rat Expression Set 230 1382946_PM_a_at - -
          Rat Expression Set 230 1392855_PM_x_at - -
          Rat Expression Set 230 1392855_x_at - -
          Rat Expression Set 230 1382946_a_at - -
          Rat Expression Set 230 1385270_s_at - -
          Rat Expression Set 230 1382947_at - -
          Affymetrix Rat Gene 1.0 ST Array 10933945 - -
          Affymetrix Rat Gene 1.0 ST Array 10727734 - -
          Rat Genome U34 Set X13905cds_at - -
          Rat Genome U34 Set X13905cds_g_at - -
          Rat Genome U34 Set rc_AA851765_at - -
          Rat Genome U34 Set rc_AI236576_at - -
          Rat Genome U34 Set rc_AI178239_at - -
          Rat Genome U34 Set X13905cds_at - -
          Rat Genome U34 Set X13905cds_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21620436 - -
          ABI Rat Genome Survey Microarray 22035126 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5756d5bff6-86w5v:80/100.66.130.62:80.
          git-commit: 27702359083d4c7b0962b032f2611a59cac079b5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0