Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504620_g1
          See other LOC100362027 GE Assays ›
          Gene Symbol:
          LOC100362027 Rpl30
          Gene Name:
          ribosomal protein L30-like
          ribosomal protein L30
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.10: 38024456 - 38024944 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 73213538 - 73216431 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01504620_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100362027
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002727709.3 XP_002727755.1 1 49 68
          Gene Symbol: Rpl30
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022699.3 NP_073190.1 1 - 2 45 68
          GenBank mRNA BC058471.1 - 1 - 2 49 68
          FQ210232.1 - 1 - 2 42 68
          FQ210454.1 - 1 - 2 41 68
          FQ210629.1 - 1 - 2 45 68
          FQ211005.1 - 1 - 2 47 68
          FQ211389.1 - 1 - 2 43 68
          FQ211694.1 - 1 - 2 42 68
          FQ211731.1 - 1 - 2 41 68
          FQ211953.1 - 1 - 2 45 68
          FQ214466.1 - 1 - 2 42 68
          FQ217830.1 - 1 - 2 42 68
          FQ217913.1 - 1 - 2 42 68
          FQ221046.1 - 1 - 2 68
          FQ221166.1 - 1 - 2 42 68
          FQ221653.1 - 1 - 2 50 68
          FQ221662.1 - 1 - 2 43 68
          FQ221836.1 - 1 - 2 42 68
          FQ222551.1 - 1 - 2 42 68
          FQ222688.1 - 1 - 2 42 68
          FQ222703.1 - 1 - 2 41 68
          FQ223191.1 - 1 - 2 41 68
          FQ223319.1 - 1 - 2 42 68
          FQ223373.1 - 1 - 2 41 68
          FQ223410.1 - 1 - 2 139 68
          FQ223604.1 - 362 68
          FQ223661.1 - 1 - 2 42 68
          FQ224433.1 - 1 - 2 44 68
          FQ224506.1 - 1 - 2 41 68
          FQ224779.1 - 1 - 2 42 68
          FQ228655.1 - 1 - 2 41 68
          FQ228991.1 - 1 - 2 41 68
          FQ229525.1 - 1 - 2 40 68
          FQ229603.1 - 642 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002958.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the BC058471.1, NM_022699.3, XM_002727709.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Endogenous Controls GE Endogenous Controls (Legacy) GE Endogenous Controls Detect Genomic DNA Rattus norvegicus Rat Endogenous Controls Inventoried Rat_Endogenous_Controls Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100362027
          Click here to find siRNAs for Gene Rpl30

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0700022899.rn.1 TC0700002958.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022898.rn.1 TC0700002958.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0