Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057124.2 | NP_476465.1 | 2 - 3 | 174 | 66 | |
XM_006229711.2 | XP_006229773.1 | 1 - 2 | 258 | 66 | ||
XM_006229712.2 | XP_006229774.1 | 2 - 3 | 1610 | 66 | ||
XM_006229713.2 | XP_006229775.1 | 2 - 3 | 327 | 66 | ||
XM_006229714.2 | XP_006229776.1 | 2 - 3 | 1274 | 66 | ||
XM_006229715.2 | XP_006229777.1 | 3 - 4 | 1757 | 66 | ||
XM_006229716.2 | XP_006229778.1 | 2 - 3 | 995 | 66 | ||
XM_006229717.2 | XP_006229779.1 | 2 - 3 | 3565 | 66 | ||
GenBank mRNA | BC072520.1 | - | 2 - 3 | 174 | 66 | |
D63665.1 | - | 3 - 4 | 398 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |