Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01513966_m1
          See other SLC43A1 GE Assays ›
          Gene Symbol:
          Slc43a1
          Gene Name:
          solute carrier family 43 (amino acid system L transporter), member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 72238796 - 72265023 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01513966_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc43a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107742.1 NP_001101212.1 13 - 14 1858 65
          GenBank mRNA BC161926.1 - 13 - 14 1819 65
          FQ226815.1 - 1 - 2 110 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000976.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          neutral amino acid transport
          L-amino acid transport
          L-alpha-amino acid transmembrane transport
          neutral amino acid transmembrane transporter activity
          L-amino acid transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P410954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31470 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76279 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P410954 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P410954 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P410954 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65931 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7239 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10837364 - -
          Affymetrix Rat Gene 1.0 ST Array 10837360 - -
          Affymetrix Rat Gene 1.0 ST Array 10837359 - -
          Affymetrix Rat Gene 1.0 ST Array 10837358 - -
          Affymetrix Rat Gene 1.0 ST Array 10837354 - -
          Affymetrix Rat Gene 1.0 ST Array 10837355 - -
          Affymetrix Rat Gene 1.0 ST Array 10837361 - -
          Affymetrix Rat Gene 1.0 ST Array 10837363 - -
          Affymetrix Rat Gene 1.0 ST Array 10837362 - -
          Affymetrix Rat Gene 1.0 ST Array 10837365 - -
          Affymetrix Rat Gene 1.0 ST Array 10837356 - -
          Affymetrix Rat Gene 1.0 ST Array 10837357 - -
          Affymetrix Rat Gene 1.0 ST Array 10837353 - -
          Affymetrix Rat Gene 1.1 ST Array 10837351 - -
          Rat Genome 230 2.0 Array 1378196_at - -
          Rat Genome 230 2.0 Array 1378196_PM_at - -
          Rat Genome 230 2.0 Array 1378865_at - -
          Rat Genome 230 2.0 Array 1378865_PM_at - -
          Affymetrix HT Rat Focus Array 1378196_at - -
          Affymetrix HT Rat Focus Array 1378196_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300007245.rn.1 TC0300000976.rn.1 RTA TC
          Rat Expression Set 230 1378196_at - -
          Rat Expression Set 230 1378196_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10837351 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22189650 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358919 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0