Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01515600_m1
See other EPB4.1 GE Assays ›
Gene Symbol:
Epb4.1
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
RGD1564762
Chromosome Location:
Chr.5: 150081727 - 150243186 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01515600_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Epb4.1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_006239085.2
XP_006239147.1
11 - 12
1936
67
XM_006239086.2
XP_006239148.1
11 - 12
1936
67
XM_006239087.2
XP_006239149.1
11 - 12
3888
67
XM_006239088.2
XP_006239150.1
11 - 12
1936
67
XM_006239089.2
XP_006239151.1
11 - 12
1936
67
XM_006239090.2
XP_006239152.1
11 - 12
1936
67
XM_006239091.2
XP_006239153.1
11 - 12
1936
67
XM_006239092.2
XP_006239154.1
11 - 12
1937
67
XM_006239093.2
XP_006239155.1
11 - 12
1772
67
XM_006239094.2
XP_006239156.1
11 - 12
1936
67
XM_006239095.1
XP_006239157.1
11 - 12
1795
67
XM_006239096.2
XP_006239158.1
11 - 12
1934
67
XM_006239097.2
XP_006239159.1
11 - 12
1934
67
XM_006239098.2
XP_006239160.1
11 - 12
1934
67
XM_006239099.2
XP_006239161.1
11 - 12
1934
67
XM_006239100.2
XP_006239162.1
11 - 12
1933
67
XM_006239101.2
XP_006239163.1
11 - 12
1935
67
XM_006239103.2
XP_006239165.1
11 - 12
1933
67
XM_006239104.2
XP_006239166.1
11 - 12
1934
67
XM_006239105.2
XP_006239167.1
11 - 12
1936
67
XM_008764184.1
XP_008762406.1
11 - 12
1933
67
XM_232771.8
XP_232771.6
11 - 12
1772
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Rattus norvegicus
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural molecule activity
protein binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix