Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515611_m1
          See other EPB4.1 GE Assays ›
          Gene Symbol:
          Epb4.1
          Gene Name:
          erythrocyte membrane protein band 4.1
          Gene Aliases:
          RGD1564762
          Chromosome Location:
          Chr.5: 150081727 - 150243186 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515611_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Epb4.1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006239085.2 XP_006239147.1 6 - 7 1199 64
          XM_006239086.2 XP_006239148.1 6 - 7 1199 64
          XM_006239087.2 XP_006239149.1 6 - 7 3151 64
          XM_006239088.2 XP_006239150.1 6 - 7 1199 64
          XM_006239089.2 XP_006239151.1 6 - 7 1199 64
          XM_006239090.2 XP_006239152.1 6 - 7 1199 64
          XM_006239091.2 XP_006239153.1 6 - 7 1199 64
          XM_006239092.2 XP_006239154.1 6 - 7 1200 64
          XM_006239093.2 XP_006239155.1 6 - 7 1035 64
          XM_006239094.2 XP_006239156.1 6 - 7 1199 64
          XM_006239095.1 XP_006239157.1 6 - 7 1058 64
          XM_006239096.2 XP_006239158.1 6 - 7 1197 64
          XM_006239097.2 XP_006239159.1 6 - 7 1197 64
          XM_006239098.2 XP_006239160.1 6 - 7 1197 64
          XM_006239099.2 XP_006239161.1 6 - 7 1197 64
          XM_006239100.2 XP_006239162.1 6 - 7 1196 64
          XM_006239101.2 XP_006239163.1 6 - 7 1198 64
          XM_006239103.2 XP_006239165.1 6 - 7 1196 64
          XM_006239104.2 XP_006239166.1 6 - 7 1197 64
          XM_006239105.2 XP_006239167.1 6 - 7 1199 64
          XM_008764184.1 XP_008762406.1 6 - 7 1196 64
          XM_008764185.1 XP_008762407.1 6 - 7 1196 64
          XM_232771.8 XP_232771.6 6 - 7 1035 64
          GenBank mRNA FQ233548.1 - 6 - 7 994 64

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          cortical actin cytoskeleton organization
          positive regulation of protein binding
          actin binding
          structural molecule activity
          protein binding
          1-phosphatidylinositol binding
          spectrin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix