Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01515839_m1
          See other SRSF10 GE Assays ›
          Gene Symbol:
          Srsf10
          Gene Name:
          serine/arginine-rich splicing factor 10
          Gene Aliases:
          Fusip1
          Chromosome Location:
          Chr.5: 154205359 - 154219550 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01515839_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srsf10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025738.1 NP_001020909.1 3 - 4 366 74
          XM_006239220.2 XP_006239282.1 3 - 4 409 74
          XM_006239221.2 XP_006239283.1 3 - 4 409 74
          XM_006239222.2 XP_006239284.1 3 - 4 407 74
          XM_006239223.2 XP_006239285.1 3 - 4 408 74
          GenBank mRNA BC098831.1 - 3 - 4 366 74
          FQ227288.1 - 3 - 4 387 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001688.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srsf10

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA splicing, via transesterification reactions
          mRNA splicing, via spliceosome
          mRNA splice site selection
          cytosolic transport
          negative regulation of mRNA splicing, via spliceosome
          nucleotide binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P408011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293670 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149383 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P408011 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P408011 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P408011 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P408011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36642 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872931 - -
          Affymetrix Rat Gene 1.0 ST Array 10934697 - -
          Affymetrix Rat Gene 1.0 ST Array 10872934 - -
          Affymetrix Rat Gene 1.0 ST Array 10872930 - -
          Affymetrix Rat Gene 1.0 ST Array 10872932 - -
          Affymetrix Rat Gene 1.0 ST Array 10872933 - -
          Affymetrix Rat Gene 1.1 ST Array 10872929 - -
          Affymetrix Rat Gene 1.1 ST Array 10934696 - -
          Rat Genome 230 2.0 Array 1380269_PM_at - -
          Rat Genome 230 2.0 Array 1393946_PM_at - -
          Rat Genome 230 2.0 Array 1383703_PM_at - -
          Rat Genome 230 2.0 Array 1392868_PM_at - -
          Rat Genome 230 2.0 Array 1386071_at - -
          Rat Genome 230 2.0 Array 1392868_at - -
          Rat Genome 230 2.0 Array 1386071_PM_at - -
          Rat Genome 230 2.0 Array 1383703_at - -
          Rat Genome 230 2.0 Array 1380269_at - -
          Rat Genome 230 2.0 Array 1393946_at - -
          Affymetrix HT Rat Focus Array 1393946_PM_at - -
          Affymetrix HT Rat Focus Array 1393946_at - -
          Affymetrix HT Rat Focus Array 1380269_at - -
          Affymetrix HT Rat Focus Array 1380269_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500012234.rn.1 TC0500001688.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012233.rn.1 TC0500001688.rn.1 RTA TC
          Rat Expression Set 230 1380269_at - -
          Rat Expression Set 230 1393946_PM_at - -
          Rat Expression Set 230 1380269_PM_at - -
          Rat Expression Set 230 1393946_at - -
          Rat Expression Set 230 1386071_at - -
          Rat Expression Set 230 1386071_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872929 - -
          Rat Genome U34 Set rc_AA859619_at - -
          Rat Genome U34 Set rc_AA859495_at - -
          Rat Genome U34 Set rc_AA859495_g_at - -
          Rat Genome U34 Set rc_AI178672_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367144 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0