Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01525640_m1
          See other DDO GE Assays ›
          Gene Symbol:
          Ddo
          Gene Name:
          D-aspartate oxidase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 48739812 - 48758896 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01525640_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109465.2 NP_001102935.1 3 - 4 339 84
          GenBank mRNA AB983592.1 - 2 - 3 279 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001742.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          aspartate catabolic process
          insemination
          grooming behavior
          D-amino acid catabolic process
          hormone metabolic process
          signaling receptor binding
          D-aspartate oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P001389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85280 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115663 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115486 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001389 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P001389 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P001389 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10830477 - -
          Affymetrix Rat Gene 1.0 ST Array 10830472 - -
          Affymetrix Rat Gene 1.0 ST Array 10830475 - -
          Affymetrix Rat Gene 1.0 ST Array 10830476 - -
          Affymetrix Rat Gene 1.0 ST Array 10830473 - -
          Affymetrix Rat Gene 1.0 ST Array 10830474 - -
          Affymetrix Rat Gene 1.1 ST Array 10830471 - -
          Affymetrix Rat Clariom S Assay PSR2000014071.rn.1 TC2000001742.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR2000014072.rn.1 TC2000001742.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10830471 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22109514 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b776c6558-ktt9x:80/100.66.134.75:80.
          git-commit: 08b7be5c3d3493be030128aff2ceec7cf49358c5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc