Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529714_m1
          See other OPN1MW GE Assays ›
          Gene Symbol:
          Opn1mw
          Gene Name:
          opsin 1 (cone pigments), medium-wave-sensitive
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 156569683 - 156589907 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529714_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Opn1mw
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053548.1 NP_446000.1 5 - 6 968 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00002640.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Opn1mw

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          visual perception
          phototransduction
          biological_process
          protein-chromophore linkage
          positive regulation of cytokinesis
          molecular_function
          G protein-coupled receptor activity
          photoreceptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P087708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806480 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138562 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087708 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P087708 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P087708 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P163221 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104517 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50378 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936032 - -
          Affymetrix Rat Gene 1.0 ST Array 10936031 - -
          Affymetrix Rat Gene 1.0 ST Array 10936030 - -
          Affymetrix Rat Gene 1.0 ST Array 10936028 - -
          Affymetrix Rat Gene 1.0 ST Array 10936027 - -
          Affymetrix Rat Gene 1.0 ST Array 10936029 - -
          Affymetrix Rat Gene 1.1 ST Array 10936026 - -
          Rat Genome 230 2.0 Array 1369913_at - -
          Rat Genome 230 2.0 Array 1369913_PM_at - -
          Affymetrix HT Rat Focus Array 1369913_at - -
          Affymetrix HT Rat Focus Array 1369913_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00016438.rn.1 TC0X00002640.rn.1 RTA TC
          Rat Expression Set 230 1369913_PM_at - -
          Rat Expression Set 230 1369913_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936026 - -
          Rat Genome U34 Set AF031528_s_at - -
          Rat Genome U34 Set AF054241mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21115822 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367704 - -
          RatRef-12 Expression BeadChip ILMN_63427 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0