Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530298_m1
          See other LOC100910288 GE Assays ›
          Gene Symbol:
          LOC100910288 LOC103689943 Marf1
          Gene Name:
          meiosis arrest female protein 1-like
          meiosis arrest female protein 1-like
          meiosis arrest female 1
          Gene Aliases:
          -
          -
          Lkap
          Chromosome Location:
          Chr.10: 1700881 - 1716659 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 28269 - 72912 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 908806 - 953481 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn01530298_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100910288
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006245699.2 XP_006245761.1 5 - 6 924 109
          Gene Symbol: LOC103689943
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006245686.2 XP_006245748.1 21 - 22 5413 109
          XM_006245687.2 XP_006245749.1 21 - 22 4297 109
          XM_006245688.1 XP_006245750.1 21 - 22 4191 109
          XM_006245689.2 XP_006245751.1 21 - 22 3802 109
          XM_008767447.1 XP_008765669.1 21 - 22 4339 109
          XM_008767448.1 XP_008765670.1 20 - 21 4846 109
          Gene Symbol: Marf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133421.2 NP_596912.1 21 - 22 4365 109
          XM_003750779.3 XP_003750827.1 21 - 22 5412 109
          XM_006245854.2 XP_006245916.1 21 - 22 4297 109
          XM_006245855.2 XP_006245917.1 21 - 22 4191 109
          XM_006245856.2 XP_006245918.1 21 - 22 3802 109
          XM_008767442.1 XP_008765664.1 20 - 21 4846 109
          GenBank mRNA AB012133.1 - 21 - 22 4365 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000012.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100910288
          Click here to find siRNAs for Gene LOC103689943
          Click here to find siRNAs for Gene Marf1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of gene expression
          double-strand break repair
          female meiotic nuclear division
          negative regulation of phosphatase activity
          oogenesis
          nucleotide binding
          nucleic acid binding
          RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1000000184.rn.1 TC1000000012.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0