Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536062_m1
          See other LOC100911034 GE Assays ›
          Gene Symbol:
          LOC100911034 Nfs1
          Gene Name:
          cysteine desulfurase, mitochondrial-like
          NFS1 nitrogen fixation 1 homolog (S. cerevisiae)
          Gene Aliases:
          -
          Nifs
          Chromosome Location:
          Chr.3: 152200322 - 152222667 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 151665813 - 151688151 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01536062_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100911034
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749609.2 XP_003749657.1 12 - 13 1341 64
          XM_006235451.2 XP_006235513.1 12 - 13 1335 64
          Gene Symbol: Nfs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053462.2 NP_445914.2 1336 64
          XM_006235365.2 XP_006235427.1 1335 64
          GenBank mRNA AF336041.1 - 1297 64
          BC089205.1 - 1302 64
          BC104699.1 - 1336 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004568.rn.1, TC0300004574.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100911034
          Click here to find siRNAs for Gene Nfs1

          Gene Ontology Categories:

          Function(s) Process(es)

          cysteine metabolic process
          iron incorporation into metallo-sulfur cluster
          Mo-molybdopterin cofactor biosynthetic process
          pyridoxal phosphate binding
          cysteine desulfurase activity
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P351159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170938 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140326 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P351159 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P351159 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P351159 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P351159 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84327 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54299 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17693 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10851160 - -
          Affymetrix Rat Gene 1.0 ST Array 10851163 - -
          Affymetrix Rat Gene 1.0 ST Array 10851159 - -
          Affymetrix Rat Gene 1.0 ST Array 10851162 - -
          Affymetrix Rat Gene 1.0 ST Array 10851161 - -
          Affymetrix Rat Gene 1.0 ST Array 10851165 - -
          Affymetrix Rat Gene 1.0 ST Array 10851158 - -
          Affymetrix Rat Gene 1.0 ST Array 10851170 - -
          Affymetrix Rat Gene 1.0 ST Array 10851172 - -
          Affymetrix Rat Gene 1.0 ST Array 10851164 - -
          Affymetrix Rat Gene 1.0 ST Array 10851168 - -
          Affymetrix Rat Gene 1.0 ST Array 10851169 - -
          Affymetrix Rat Gene 1.0 ST Array 10851167 - -
          Affymetrix Rat Gene 1.0 ST Array 10851166 - -
          Affymetrix Rat Gene 1.0 ST Array 10851171 - -
          Affymetrix Rat Gene 1.1 ST Array 10851157 - -
          Rat Genome 230 2.0 Array 1387859_PM_at - -
          Rat Genome 230 2.0 Array 1387859_at - -
          Affymetrix HT Rat Focus Array 1387859_PM_at - -
          Affymetrix HT Rat Focus Array 1387859_at - -
          Affymetrix Rat Clariom S Assay PSR0300035267.rn.1 TC0300004574.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035116.rn.1 TC0300004568.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035115.rn.1 TC0300004568.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035117.rn.1 TC0300004568.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035118.rn.1 TC0300004568.rn.1 RTA TC
          Rat Expression Set 230 1387859_at - -
          Rat Expression Set 230 1387859_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10851157 - -
          Rat Genome U34 Set rc_AI235312_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22064871 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040171 - -
          RatRef-12 Expression BeadChip ILMN_68857 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix