Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536150_m1
          See other CST7 GE Assays ›
          Gene Symbol:
          Cst7
          Gene Name:
          cystatin F (leukocystatin)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 146363915 - 146374297 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536150_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cst7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106523.1 NP_001099993.1 2 - 3 386 64
          XM_006235213.2 XP_006235275.1 3 - 4 1659 64
          XM_008762305.1 XP_008760527.1 3 - 4 1659 64
          XM_008762306.1 XP_008760528.1 3 - 4 1656 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001985.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of endopeptidase activity
          cysteine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P131693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531773 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131693 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131693 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P131693 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P832728 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70413 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840678 - -
          Affymetrix Rat Gene 1.0 ST Array 10840675 - -
          Affymetrix Rat Gene 1.0 ST Array 10840676 - -
          Affymetrix Rat Gene 1.0 ST Array 10840679 - -
          Affymetrix Rat Gene 1.0 ST Array 10840677 - -
          Affymetrix Rat Gene 1.1 ST Array 10840673 - -
          Rat Genome 230 2.0 Array 1382622_at - -
          Rat Genome 230 2.0 Array 1382622_PM_at - -
          Affymetrix HT Rat Focus Array 1382622_PM_at - -
          Affymetrix HT Rat Focus Array 1382622_at - -
          Affymetrix Rat Clariom S Assay PSR0300014142.rn.1 TC0300001985.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300014144.rn.1 TC0300001985.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300014138.rn.1 TC0300001985.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300014143.rn.1 TC0300001985.rn.1 RTA TC
          Rat Expression Set 230 1382622_PM_at - -
          Rat Expression Set 230 1382622_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840673 - -
          Rat Genome U34 Set rc_AI170805_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21121605 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365619 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0