Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536832_m1
          See other MCM5 GE Assays ›
          Gene Symbol:
          Mcm5
          Gene Name:
          minichromosome maintenance complex component 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 14523495 - 14561281 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536832_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mcm5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106170.1 NP_001099640.1 13 - 14 1920 59
          XM_008772283.1 XP_008770505.1 15 - 16 2054 59
          GenBank mRNA BC166442.1 - 15 - 16 1995 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000146.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication initiation
          DNA duplex unwinding
          cell division
          DNA binding
          DNA helicase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P014774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13493 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014774 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P014774 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P014774 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P546123 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94040 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806145 - -
          Affymetrix Rat Gene 1.0 ST Array 10806132 - -
          Affymetrix Rat Gene 1.0 ST Array 10806146 - -
          Affymetrix Rat Gene 1.0 ST Array 10806134 - -
          Affymetrix Rat Gene 1.0 ST Array 10806137 - -
          Affymetrix Rat Gene 1.0 ST Array 10806133 - -
          Affymetrix Rat Gene 1.0 ST Array 10806140 - -
          Affymetrix Rat Gene 1.0 ST Array 10806135 - -
          Affymetrix Rat Gene 1.0 ST Array 10806138 - -
          Affymetrix Rat Gene 1.0 ST Array 10806144 - -
          Affymetrix Rat Gene 1.0 ST Array 10806136 - -
          Affymetrix Rat Gene 1.0 ST Array 10806143 - -
          Affymetrix Rat Gene 1.0 ST Array 10806139 - -
          Affymetrix Rat Gene 1.0 ST Array 10806131 - -
          Affymetrix Rat Gene 1.0 ST Array 10806142 - -
          Affymetrix Rat Gene 1.0 ST Array 10806141 - -
          Affymetrix Rat Gene 1.1 ST Array 10806129 - -
          Rat Genome 230 2.0 Array 1376055_at - -
          Rat Genome 230 2.0 Array 1376055_PM_at - -
          Affymetrix HT Rat Focus Array 1376055_at - -
          Affymetrix HT Rat Focus Array 1376055_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900001170.rn.1 TC1900000146.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001167.rn.1 TC1900000146.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001169.rn.1 TC1900000146.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001166.rn.1 TC1900000146.rn.1 RTA TC
          Rat Expression Set 230 1376055_PM_at - -
          Rat Expression Set 230 1376055_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806129 - -
          Rat Genome U34 Set rc_AA859768_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21884767 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2038852 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040160 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0