Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01646721_s1
          See other KCNJ16 GE Assays ›
          Gene Symbol:
          Kcnj16
          Gene Name:
          potassium channel, inwardly rectifying subfamily J, member 16
          Gene Aliases:
          Kir5.1
          Chromosome Location:
          Chr.10: 99330894 - 99391551 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01646721_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnj16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053314.2 NP_445766.2 1 1259 114
          XM_006247568.2 XP_006247630.1 2 1412 114
          XM_006247569.2 XP_006247631.1 2 1499 114
          XM_006247570.2 XP_006247632.1 2 2077 114
          XM_008768347.1 XP_008766569.1 2 1503 114
          GenBank mRNA BC078776.1 - 1 1259 114

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnj16

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          regulation of ion transmembrane transport
          potassium ion transmembrane transport
          potassium ion import across plasma membrane
          inward rectifier potassium channel activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P833096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P833096 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P833096 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P833096 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P833096 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54961 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739352 - -
          Affymetrix Rat Gene 1.1 ST Array 10739351 - -
          Rat Genome 230 2.0 Array 1373991_at - -
          Rat Genome 230 2.0 Array 1373991_PM_at - -
          Rat Genome 230 2.0 Array 1370076_at - -
          Rat Genome 230 2.0 Array 1370076_PM_at - -
          Affymetrix HT Rat Focus Array 1370076_PM_at - -
          Affymetrix HT Rat Focus Array 1370076_at - -
          Rat Expression Set 230 1370076_PM_at - -
          Rat Expression Set 230 1370076_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739351 - -
          Rat Genome U34 Set rc_AA892312_at - -
          Rat Genome U34 Set X83581cds_at - -
          Rat Genome U34 Set X83581cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20711171 - -
          ABI Rat Genome Survey Microarray 21749912 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370340 - -
          RatRef-12 Expression BeadChip ILMN_48168 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0