Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01647685_g1
          See other NME1 GE Assays ›
          Gene Symbol:
          Nme1
          Gene Name:
          NME/NM23 nucleoside diphosphate kinase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 81657153 - 81666523 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01647685_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nme1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138548.2 NP_612557.1 3 - 4 343 58
          GenBank mRNA D13374.1 - 3 - 4 344 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003576.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nme1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of myeloid leukocyte differentiation
          purine nucleotide metabolic process
          nucleoside diphosphate phosphorylation
          GTP biosynthetic process
          pyrimidine nucleotide metabolic process
          UTP biosynthetic process
          CTP biosynthetic process
          DNA catabolic process
          endocytosis
          integrin-mediated signaling pathway
          lactation
          nucleoside triphosphate biosynthetic process
          response to xenobiotic stimulus
          negative regulation of gene expression
          positive regulation of neuron projection development
          response to amine
          hippocampus development
          mammary gland development
          response to testosterone
          regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of DNA binding
          positive regulation of keratinocyte differentiation
          positive regulation of transcription by RNA polymerase II
          positive regulation of epithelial cell proliferation
          response to cAMP
          cellular response to glucose stimulus
          cellular response to fatty acid
          cellular response to xenobiotic stimulus
          magnesium ion binding
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          single-stranded DNA binding
          RNA binding
          deoxyribonuclease activity
          nucleoside diphosphate kinase activity
          protein binding
          ATP binding
          GTP binding
          intermediate filament binding
          enzyme binding
          protein kinase binding
          identical protein binding
          gamma-tubulin binding
          ribosomal small subunit binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P407138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161211 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P407138 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P407138 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P407138 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P407138 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700245761 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51537 - -
          Agilent Rat cDNA Microarray (G4105A) 700245761 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10746278 - -
          Affymetrix Rat Gene 1.0 ST Array 10746277 - -
          Affymetrix Rat Gene 1.0 ST Array 10746279 - -
          Affymetrix Rat Gene 1.0 ST Array 10746280 - -
          Affymetrix Rat Gene 1.1 ST Array 10746276 - -
          Rat Genome 230 2.0 Array 1370295_PM_at - -
          Rat Genome 230 2.0 Array 1370295_at - -
          Affymetrix HT Rat Focus Array 1370295_at - -
          Affymetrix HT Rat Focus Array 1370295_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000031813.rn.1 TC1000003576.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031811.rn.1 TC1000003576.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000031812.rn.1 TC1000003576.rn.1 RTA TC
          Rat Expression Set 230 1370295_at - -
          Rat Expression Set 230 1370295_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10746276 - -
          Rat Genome U34 Set rc_AI233173_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22137585 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369051 - -
          RatRef-12 Expression BeadChip ILMN_62560 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix