Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01750092_g1
          See other USP29 GE Assays ›
          Gene Symbol:
          Usp29
          Gene Name:
          ubiquitin specific peptidase 29
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 69988673 - 70042015 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01750092_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Usp29
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108465.1 NP_001101935.1 5 - 6 524 103
          XM_006228185.2 XP_006228247.1 5 - 6 524 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004737.rn.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001108465.1, XM_006228185.2 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein deubiquitination
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of proteasomal protein catabolic process
          cysteine-type endopeptidase activity
          thiol-dependent deubiquitinase

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P550567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150746 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65959 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178738 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134303 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550567 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P550567 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P550567 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P550567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47023 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10718665 - -
          Affymetrix Rat Gene 1.1 ST Array 10718664 - -
          Rat Genome 230 2.0 Array 1395054_at - -
          Rat Genome 230 2.0 Array 1395054_PM_at - -
          Affymetrix HT Rat Focus Array 1395054_at - -
          Affymetrix HT Rat Focus Array 1395054_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100037314.rn.1 TC0100004737.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100037315.rn.1 TC0100004737.rn.1 RTA TC
          Rat Expression Set 230 1395054_at - -
          Rat Expression Set 230 1395054_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10718664 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21854097 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650518 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0