Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01755556_m1
          See other MIB1 GE Assays ›
          Gene Symbol:
          Mib1
          Gene Name:
          mindbomb E3 ubiquitin protein ligase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 1970914 - 2094920 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01755556_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mib1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107405.1 NP_001100875.1 9 - 10 2053 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000025.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          in utero embryonic development
          somitogenesis
          neural tube formation
          heart looping
          endocytosis
          Notch signaling pathway
          heart development
          biological_process
          protein ubiquitination
          negative regulation of neuron differentiation
          positive regulation of endocytosis
          molecular_function
          ubiquitin-protein transferase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P398492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491314 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150871 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49424 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172451 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110632 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398492 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P398492 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P398492 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P398492 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47878 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3661 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65369 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85033 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5926 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65179 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88622 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10800154 - -
          Affymetrix Rat Gene 1.0 ST Array 10800148 - -
          Affymetrix Rat Gene 1.0 ST Array 10800151 - -
          Affymetrix Rat Gene 1.0 ST Array 10800162 - -
          Affymetrix Rat Gene 1.0 ST Array 10800144 - -
          Affymetrix Rat Gene 1.0 ST Array 10800163 - -
          Affymetrix Rat Gene 1.0 ST Array 10800153 - -
          Affymetrix Rat Gene 1.0 ST Array 10800152 - -
          Affymetrix Rat Gene 1.0 ST Array 10800150 - -
          Affymetrix Rat Gene 1.0 ST Array 10800146 - -
          Affymetrix Rat Gene 1.0 ST Array 10800159 - -
          Affymetrix Rat Gene 1.0 ST Array 10800145 - -
          Affymetrix Rat Gene 1.0 ST Array 10800160 - -
          Affymetrix Rat Gene 1.0 ST Array 10800157 - -
          Affymetrix Rat Gene 1.0 ST Array 10800147 - -
          Affymetrix Rat Gene 1.0 ST Array 10800156 - -
          Affymetrix Rat Gene 1.0 ST Array 10800161 - -
          Affymetrix Rat Gene 1.0 ST Array 10800143 - -
          Affymetrix Rat Gene 1.0 ST Array 10800155 - -
          Affymetrix Rat Gene 1.0 ST Array 10800149 - -
          Affymetrix Rat Gene 1.0 ST Array 10800158 - -
          Affymetrix Rat Gene 1.1 ST Array 10800142 - -
          Affymetrix Rat Clariom S Assay PSR1800000226.rn.1 TC1800000025.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10800142 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21655529 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359374 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0