Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01755753_m1
          See other PGRMC2 GE Assays ›
          Gene Symbol:
          Pgrmc2
          Gene Name:
          progesterone receptor membrane component 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 127986106 - 128002005 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01755753_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pgrmc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008374.1 NP_001008375.1 2 - 3 551 80
          GenBank mRNA BC083571.1 - 2 - 3 551 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003575.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pgrmc2

          Gene Ontology Categories:

          Process(es)

          steroid binding
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P126080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43125 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P126080 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P126080 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P126080 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P228839 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P492189 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103595 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25366 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822984 - -
          Affymetrix Rat Gene 1.0 ST Array 10822986 - -
          Affymetrix Rat Gene 1.0 ST Array 10822985 - -
          Affymetrix Rat Gene 1.1 ST Array 10822983 - -
          Rat Genome 230 2.0 Array 1388531_at - -
          Rat Genome 230 2.0 Array 1388531_PM_at - -
          Rat Genome 230 2.0 Array 1389169_PM_at - -
          Rat Genome 230 2.0 Array 1389169_at - -
          Rat Genome 230 2.0 Array 1379427_at - -
          Rat Genome 230 2.0 Array 1379427_PM_at - -
          Affymetrix HT Rat Focus Array 1379427_at - -
          Affymetrix HT Rat Focus Array 1379427_PM_at - -
          Affymetrix HT Rat Focus Array 1389169_at - -
          Affymetrix HT Rat Focus Array 1389169_PM_at - -
          Affymetrix HT Rat Focus Array 1388531_at - -
          Affymetrix HT Rat Focus Array 1388531_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200024183.rn.1 TC0200003575.rn.1 RTA TC
          Rat Expression Set 230 1389169_PM_at - -
          Rat Expression Set 230 1388531_PM_at - -
          Rat Expression Set 230 1388531_at - -
          Rat Expression Set 230 1389169_at - -
          Rat Expression Set 230 1379427_at - -
          Rat Expression Set 230 1379427_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822983 - -
          Rat Genome U34 Set rc_AI137488_at - -
          Rat Genome U34 Set rc_AI102871_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21947230 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360328 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530371 - -
          RatRef-12 Expression BeadChip ILMN_160794 - -
          RatRef-12 Expression BeadChip ILMN_55596 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix