Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01756109_m1
          See other ABHD6 GE Assays ›
          Gene Symbol:
          Abhd6
          Gene Name:
          abhydrolase domain containing 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 18647905 - 18695133 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01756109_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abhd6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007680.1 NP_001007681.1 8 - 9 906 95
          XM_006251766.2 XP_006251828.1 8 - 9 2185 95
          XM_006251767.2 XP_006251829.1 8 - 9 1717 95
          XM_006251768.2 XP_006251830.1 8 - 9 882 95
          GenBank mRNA BC083826.1 - 8 - 9 906 95
          FQ219067.1 - 3 - 4 184 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001419.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abhd6

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell migration
          acylglycerol catabolic process
          long-term synaptic depression
          regulation of endocannabinoid signaling pathway
          acylglycerol lipase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P289862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967388 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11958 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88587 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64003 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P289862 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P289862 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P289862 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P289862 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P128447 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83619 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26591 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34700 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10782698 - -
          Affymetrix Rat Gene 1.0 ST Array 10782696 - -
          Affymetrix Rat Gene 1.0 ST Array 10782702 - -
          Affymetrix Rat Gene 1.0 ST Array 10782697 - -
          Affymetrix Rat Gene 1.0 ST Array 10782699 - -
          Affymetrix Rat Gene 1.0 ST Array 10782700 - -
          Affymetrix Rat Gene 1.0 ST Array 10782701 - -
          Affymetrix Rat Gene 1.0 ST Array 10782703 - -
          Affymetrix Rat Gene 1.1 ST Array 10782695 - -
          Rat Genome 230 2.0 Array 1390155_PM_at - -
          Rat Genome 230 2.0 Array 1390155_at - -
          Affymetrix HT Rat Focus Array 1390155_at - -
          Affymetrix HT Rat Focus Array 1390155_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500009019.rn.1 TC1500001419.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500009020.rn.1 TC1500001419.rn.1 RTA TC
          Rat Expression Set 230 1390155_PM_at - -
          Rat Expression Set 230 1390155_at - -
          Affymetrix Rat Gene 1.0 ST Array 10782695 - -
          Rat Genome U34 Set rc_AA924793_at - -
          Rat Genome U34 Set rc_AI235569_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21540712 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364746 - -
          RatRef-12 Expression BeadChip ILMN_51332 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0