Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108059.3 | NP_001101529.1 | 5 - 6 | 1289 | 56 | |
XM_006256614.2 | XP_006256676.1 | 4 - 5 | 2271 | 56 | ||
GenBank mRNA | EF429098.1 | - | 3 - 4 | 1087 | 56 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |