Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01757314_g1
          See other LOC100911260 GE Assays ›
          Gene Symbol:
          LOC100911260 Hcfc1
          Gene Name:
          host cell factor 1-like
          host cell factor C1
          Gene Aliases:
          -
          RGD1563804
          Chromosome Location:
          Chr.X: 156759369 - 156771130 on Build Rnor_6.0
          Chromosome Location:
          Chr.X: 156812057 - 156837227 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01757314_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100911260
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006229638.1 XP_006229700.1 13 - 14 4016 58
          Gene Symbol: Hcfc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001139507.1 NP_001132979.1 24 - 25 7153 58
          XM_006229593.2 XP_006229655.1 24 - 25 7267 58
          XM_006229594.2 XP_006229656.1 24 - 25 7263 58
          XM_006229596.2 XP_006229658.1 24 - 25 7132 58
          XM_006229597.1 XP_006229659.1 24 - 25 7148 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001304.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          positive regulation of gene expression
          release from viral latency
          regulation of protein-containing complex assembly
          histone H4-K5 acetylation
          histone H4-K8 acetylation
          histone H4-K16 acetylation
          positive regulation of transcription by RNA polymerase II
          protein stabilization
          cellular response to organic cyclic compound
          molecular_function
          chromatin binding
          transcription coactivator activity
          identical protein binding
          histone acetyltransferase activity (H4-K5 specific)
          histone acetyltransferase activity (H4-K8 specific)
          histone acetyltransferase activity (H4-K16 specific)

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067310 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72300 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119800 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067310 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067305 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067305 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067310 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067310 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067305 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P552807 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10940289 - -
          Affymetrix Rat Gene 1.0 ST Array 10940293 - -
          Affymetrix Rat Gene 1.0 ST Array 10940275 - -
          Affymetrix Rat Gene 1.0 ST Array 10940282 - -
          Affymetrix Rat Gene 1.0 ST Array 10940284 - -
          Affymetrix Rat Gene 1.0 ST Array 10940287 - -
          Affymetrix Rat Gene 1.0 ST Array 10940295 - -
          Affymetrix Rat Gene 1.0 ST Array 10940278 - -
          Affymetrix Rat Gene 1.0 ST Array 10940273 - -
          Affymetrix Rat Gene 1.0 ST Array 10940281 - -
          Affymetrix Rat Gene 1.0 ST Array 10940276 - -
          Affymetrix Rat Gene 1.0 ST Array 10940277 - -
          Affymetrix Rat Gene 1.0 ST Array 10940291 - -
          Affymetrix Rat Gene 1.0 ST Array 10940274 - -
          Affymetrix Rat Gene 1.0 ST Array 10940272 - -
          Affymetrix Rat Gene 1.0 ST Array 10940283 - -
          Affymetrix Rat Gene 1.0 ST Array 10940290 - -
          Affymetrix Rat Gene 1.0 ST Array 10940285 - -
          Affymetrix Rat Gene 1.0 ST Array 10940280 - -
          Affymetrix Rat Gene 1.0 ST Array 10940292 - -
          Affymetrix Rat Gene 1.0 ST Array 10940288 - -
          Affymetrix Rat Gene 1.0 ST Array 10940294 - -
          Affymetrix Rat Gene 1.0 ST Array 10940279 - -
          Affymetrix Rat Gene 1.0 ST Array 10940286 - -
          Affymetrix Rat Gene 1.1 ST Array 10940271 - -
          Rat Genome 230 2.0 Array 1372530_PM_at - -
          Rat Genome 230 2.0 Array 1372530_at - -
          Affymetrix HT Rat Focus Array 1372530_PM_at - -
          Affymetrix HT Rat Focus Array 1372530_at - -
          Affymetrix Rat Clariom S Assay PSR0X00007914.rn.1 TC0X00001304.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007913.rn.1 TC0X00001304.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007912.rn.1 TC0X00001304.rn.1 RTA TC
          Rat Expression Set 230 1372530_PM_at - -
          Rat Expression Set 230 1372530_at - -
          Rat Genome U34 Set rc_AA944190_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350267 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix