Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01765889_gH
          See other RGD1559459 GE Assays ›
          Gene Symbol:
          RGD1559459 Ugt2b10
          Gene Name:
          similar to Expressed sequence AI788959
          UDP glucuronosyltransferase 2 family, polypeptide B10
          Gene Aliases:
          -
          Ugt2b34
          Chromosome Location:
          Chr.14: 22932464 - 22955663 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 22806132 - 22825444 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01765889_gH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1559459
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751344.3 XP_003751392.1 5 - 6 1281 86
          XM_006250809.2 XP_006250871.1 6 - 7 1345 86
          XM_008770101.1 XP_008768323.1 4 - 5 544 86
          Gene Symbol: Ugt2b10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191676.1 NP_001178605.1 4 - 5 1100 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002464.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          flavonoid biosynthetic process
          cellular response to hormone stimulus
          flavonoid glucuronidation
          biological_process
          glucuronosyltransferase activity
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P128396 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P072638 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072638 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P128396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P128396 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P072638 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P128396 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P072638 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P128396 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10772001 - -
          Affymetrix Rat Gene 1.0 ST Array 10772000 - -
          Affymetrix Rat Gene 1.0 ST Array 10771999 - -
          Affymetrix Rat Clariom S Assay PSR1400018291.rn.1 TC1400002464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018292.rn.1 TC1400002464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018297.rn.1 TC1400002464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018294.rn.1 TC1400002464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018295.rn.1 TC1400002464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018296.rn.1 TC1400002464.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400018293.rn.1 TC1400002464.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10771998 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366231 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364802 - -
          RatRef-12 Expression BeadChip ILMN_62515 - -
          RatRef-12 Expression BeadChip ILMN_64654 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0