Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01767656_m1
          See other CCDC134 GE Assays ›
          Gene Symbol:
          Ccdc134
          Gene Name:
          coiled-coil domain containing 134
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 123362469 - 123376930 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01767656_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc134
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024355.1 NP_001019526.1 5 - 6 768 117
          XM_006242113.2 XP_006242175.1 6 - 7 681 117
          GenBank mRNA BC088206.1 - 5 - 6 768 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001519.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc134

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P338068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 477689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927929 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131659 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P338068 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P338068 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P338068 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P338068 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96153 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898086 - -
          Affymetrix Rat Gene 1.0 ST Array 10898090 - -
          Affymetrix Rat Gene 1.0 ST Array 10898089 - -
          Affymetrix Rat Gene 1.0 ST Array 10898087 - -
          Affymetrix Rat Gene 1.0 ST Array 10898085 - -
          Affymetrix Rat Gene 1.0 ST Array 10898084 - -
          Affymetrix Rat Gene 1.0 ST Array 10898088 - -
          Affymetrix Rat Gene 1.0 ST Array 10898083 - -
          Affymetrix Rat Gene 1.1 ST Array 10898082 - -
          Rat Genome 230 2.0 Array 1376281_PM_at - -
          Rat Genome 230 2.0 Array 1376281_at - -
          Affymetrix Rat Clariom S Assay PSR0700011619.rn.1 TC0700001519.rn.1 RTA TC
          Rat Expression Set 230 1376281_at - -
          Rat Expression Set 230 1376281_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898082 - -
          Rat Genome U34 Set rc_AI070946_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21835372 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373552 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix