Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01768233_m1
          See other SLC22A22 GE Assays ›
          Gene Symbol:
          Slc22a22
          Gene Name:
          solute carrier family 22 (organic cation transporter), member 22
          Gene Aliases:
          RGD1304605, oat-pg
          Chromosome Location:
          Chr.7: 97042176 - 97071984 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01768233_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc22a22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013947.1 NP_001013969.1 9 - 10 1448 93
          XM_008765502.1 XP_008763724.1 9 - 10 1416 93
          GenBank mRNA AB559554.1 - 9 - 10 1445 93
          BC081982.1 - 9 - 10 1448 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003157.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc22a22

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          transmembrane transport
          sodium-independent prostaglandin transport
          prostaglandin receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P469016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80451 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37427 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P469016 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P469016 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P469016 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P469016 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3785 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61798 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903841 - -
          Affymetrix Rat Gene 1.0 ST Array 10903833 - -
          Affymetrix Rat Gene 1.0 ST Array 10903836 - -
          Affymetrix Rat Gene 1.0 ST Array 10903838 - -
          Affymetrix Rat Gene 1.0 ST Array 10903834 - -
          Affymetrix Rat Gene 1.0 ST Array 10903835 - -
          Affymetrix Rat Gene 1.0 ST Array 10903832 - -
          Affymetrix Rat Gene 1.0 ST Array 10903839 - -
          Affymetrix Rat Gene 1.0 ST Array 10903837 - -
          Affymetrix Rat Gene 1.0 ST Array 10903840 - -
          Affymetrix Rat Gene 1.1 ST Array 10903831 - -
          Affymetrix Rat Clariom S Assay PSR0700024154.rn.1 TC0700003157.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10903831 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371326 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix