Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01789993_u1
          See other RPS12 GE Assays ›
          Gene Symbol:
          Rps12 LOC100360573
          Gene Name:
          ribosomal protein S12
          ribosomal protein S12-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.1: 22758214 - 22760370 on Build Rnor_6.0
          Chromosome Location:
          Chr.19: 38619682 - 38620321 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01789993_u1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031709.3 NP_113897.2 5 390 160
          GenBank mRNA BC058460.1 - 5 394 160
          FQ210032.1 - 5 355 160
          FQ212402.1 - 5 353 160
          FQ212636.1 - 5 354 160
          FQ217474.1 - 5 354 160
          FQ221108.1 - 5 354 160
          FQ221199.1 - 5 354 160
          FQ221263.1 - 5 354 160
          FQ221353.1 - 5 356 160
          FQ221368.1 - 5 354 160
          FQ221420.1 - 5 354 160
          FQ221550.1 - 5 355 160
          FQ221711.1 - 5 356 160
          FQ222180.1 - 5 355 160
          FQ222260.1 - 5 354 160
          FQ222326.1 - 5 352 160
          FQ222422.1 - 5 354 160
          FQ222486.1 - 5 354 160
          FQ222505.1 - 5 355 160
          FQ222589.1 - 5 354 160
          FQ222591.1 - 5 354 160
          FQ222620.1 - 5 354 160
          FQ222621.1 - 5 354 160
          FQ222742.1 - 5 357 160
          FQ222863.1 - 5 354 160
          FQ222888.1 - 5 354 160
          FQ222895.1 - 5 354 160
          FQ223029.1 - 5 355 160
          FQ223134.1 - 5 355 160
          FQ223305.1 - 5 357 160
          FQ223374.1 - 5 357 160
          FQ223466.1 - 5 355 160
          FQ224074.1 - 5 355 160
          FQ226180.1 - 5 354 160
          FQ228598.1 - 5 354 160
          FQ228992.1 - 5 355 160
          M18547.1 - 5 353 160
          Gene Symbol: LOC100360573
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_002728597.4 XP_002728643.1 2 354 160

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000249.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps12
          Click here to find siRNAs for Gene LOC100360573

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          biological_process
          molecular_function
          RNA binding
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P104950 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P047798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55416 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96788 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104950 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047798 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P047798 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P104950 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P047798 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P104950 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P283546 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P283544 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96807 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10702288 - -
          Affymetrix Rat Gene 1.0 ST Array 10702287 - -
          Affymetrix Rat Gene 1.0 ST Array 10702292 - -
          Affymetrix Rat Gene 1.0 ST Array 10702289 - -
          Affymetrix Rat Gene 1.0 ST Array 10702290 - -
          Affymetrix Rat Gene 1.0 ST Array 10807516 - -
          Affymetrix Rat Gene 1.0 ST Array 10702291 - -
          Affymetrix Rat Gene 1.0 ST Array 10807515 - -
          Affymetrix Rat Gene 1.0 ST Array 10839877 - -
          Affymetrix Rat Gene 1.0 ST Array 10716086 - -
          Affymetrix Rat Gene 1.1 ST Array 10702286 - -
          Affymetrix Rat Gene 1.1 ST Array 10716085 - -
          Affymetrix Rat Gene 1.1 ST Array 10807514 - -
          Affymetrix Rat Gene 1.1 ST Array 10839876 - -
          Rat Genome 230 2.0 Array 1367640_PM_at - -
          Rat Genome 230 2.0 Array 1367640_at - -
          Affymetrix HT Rat Focus Array 1367640_PM_at - -
          Affymetrix HT Rat Focus Array 1367640_at - -
          Affymetrix Rat Clariom S Assay PSR0100001410.rn.1 TC0100000249.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100001411.rn.1 TC0100000249.rn.1 RTA TC
          Rat Expression Set 230 1367640_PM_at - -
          Rat Expression Set 230 1367640_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839876 - -
          Affymetrix Rat Gene 1.0 ST Array 10716085 - -
          Affymetrix Rat Gene 1.0 ST Array 10807514 - -
          Affymetrix Rat Gene 1.0 ST Array 10702286 - -
          Rat Genome U34 Set rc_AI103074_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21899948 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356995 - -
          RatRef-12 Expression BeadChip ILMN_51122 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix