Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn02134053_s1
          See other ACKR3 GE Assays ›
          Gene Symbol:
          Ackr3
          Gene Name:
          atypical chemokine receptor 3
          Gene Aliases:
          Cmkor1, Cxcr7, Rdc1
          Chromosome Location:
          Chr.9: 97355879 - 97367455 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn02134053_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ackr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053352.1 NP_445804.1 2 1849 108
          XM_006245479.2 XP_006245541.1 3 109
          XM_008767299.1 XP_008765521.1 2 109
          XM_008767300.1 XP_008765522.1 3 109
          GenBank mRNA AF118816.1 - 2 1849 108

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ackr3

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          vasculogenesis
          chemotaxis
          cell adhesion
          G protein-coupled receptor signaling pathway
          receptor internalization
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          scavenger receptor activity
          coreceptor activity
          C-X-C chemokine receptor activity
          C-X-C chemokine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P252483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828053 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32916 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P252483 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P252483 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P252483 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P252483 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701718874 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88288 - -
          Agilent Rat cDNA Microarray (G4105A) 701718874 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10925292 - -
          Affymetrix Rat Gene 1.0 ST Array 10925293 - -
          Affymetrix Rat Gene 1.1 ST Array 10925291 - -
          Rat Genome 230 2.0 Array 1367940_PM_at - -
          Rat Genome 230 2.0 Array 1367940_at - -
          Affymetrix HT Rat Focus Array 1367940_PM_at - -
          Affymetrix HT Rat Focus Array 1367940_at - -
          Rat Expression Set 230 1367940_at - -
          Rat Expression Set 230 1367940_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10925291 - -
          Rat Genome U34 Set AJ010828_at - -
          Rat Genome U34 Set rc_AA997721_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22244986 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361010 - -
          RatRef-12 Expression BeadChip ILMN_48673 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix