Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03682747_m1
          See other RGD1559747 GE Assays ›
          Gene Symbol:
          RGD1559747
          Gene Name:
          similar to Zinc finger and SCAN domain containing protein 2 (Zinc finger protein 29)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 78263126 - 78269064 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn03682747_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1559747
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027235.1 3 - 4 1175 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CK600316.1 - 92 Chr. - - [-] on Build 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger protein 775 Zfp775 RGD1304910,
          Znf775
          -7.0kb Sense Chr. 4 - 78240325 - 78258545 on NCBI Build Rnor_6.0
          Downstream GTPase,
          IMAP family member 8
          Gimap8 IanT 14.3kb Sense Chr. 4 - 78283357 - 78294838 on NCBI Build Rnor_6.0

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000858.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P092334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132730 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092334 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P092334 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P092334 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P154498 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855373 - -
          Affymetrix Rat Gene 1.1 ST Array 10855371 - -
          Rat Genome 230 2.0 Array 1391189_PM_a_at - -
          Rat Genome 230 2.0 Array 1378953_PM_at - -
          Rat Genome 230 2.0 Array 1378953_at - -
          Rat Genome 230 2.0 Array 1391189_a_at - -
          Affymetrix HT Rat Focus Array 1391189_PM_a_at - -
          Affymetrix HT Rat Focus Array 1378953_at - -
          Affymetrix HT Rat Focus Array 1391189_a_at - -
          Affymetrix HT Rat Focus Array 1378953_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400006134.rn.1 TC0400000858.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006133.rn.1 TC0400000858.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400006135.rn.1 TC0400000858.rn.1 RTA TC
          Rat Expression Set 230 1391189_a_at - -
          Rat Expression Set 230 1391189_PM_a_at - -
          Rat Expression Set 230 1378953_PM_at - -
          Rat Expression Set 230 1378953_at - -
          Rat Genome U34 Set rc_AA892659_at - -
          Rat Genome U34 Set rc_AA892659_g_at - -
          Rat Genome U34 Set rc_AI232306_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21951021 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1348847 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0