Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04219387_g1
          See other MGP GE Assays ›
          Gene Symbol:
          Mgp
          Gene Name:
          matrix Gla protein
          Gene Aliases:
          Mglap
          Chromosome Location:
          Chr.4: 170856783 - 170860105 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04219387_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mgp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012862.1 NP_036994.1 3 - 4 180 56
          GenBank mRNA BC086394.1 - 3 - 4 212 56
          FQ224403.1 - 3 - 4 237 56
          J03026.1 - 3 - 4 180 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400003973.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mgp

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          response to nutrient
          response to mechanical stimulus
          response to hormone
          response to organic substance
          cell differentiation
          lung development
          regulation of bone mineralization
          branching morphogenesis of an epithelial tube
          cartilage development
          response to glucocorticoid
          response to calcium ion
          protein-containing complex assembly
          calcium ion binding
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P588944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106250 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P588944 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P588944 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P588944 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P588944 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700821975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90487 - -
          Agilent Rat cDNA Microarray (G4105A) 700821975 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10866516 - -
          Affymetrix Rat Gene 1.0 ST Array 10866513 - -
          Affymetrix Rat Gene 1.0 ST Array 10866514 - -
          Affymetrix Rat Gene 1.0 ST Array 10866515 - -
          Affymetrix Rat Gene 1.1 ST Array 10866512 - -
          Rat Genome 230 2.0 Array 1392578_PM_at - -
          Rat Genome 230 2.0 Array 1392578_at - -
          Rat Genome 230 2.0 Array 1367568_a_at - -
          Rat Genome 230 2.0 Array 1367568_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367568_a_at - -
          Affymetrix HT Rat Focus Array 1367568_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0400028210.rn.1 TC0400003973.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400028209.rn.1 TC0400003973.rn.1 RTA TC
          Rat Expression Set 230 1367568_a_at - -
          Rat Expression Set 230 1367568_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10866512 - -
          Rat Genome U34 Set rc_AI012030_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351917 - -
          RatRef-12 Expression BeadChip ILMN_51685 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0