Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04222875_gH
          See other RRLT GE Assays ›
          Gene Symbol:
          Rrlt Raet1d Raet1l
          Gene Name:
          similar to retinoic acid early transcript 1L
          retinoic acid early transcript delta
          retinoic acid early transcript 1L
          Gene Aliases:
          -
          RGD1565376
          -
          Chromosome Location:
          Chr.1: 1437668 - 1441961 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 1470800 - 1475152 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 114233896 - 114234243 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Rn04222875_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rrlt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161691.1 NP_001155163.1 578 93
          GenBank mRNA FJ971664.1 - 578 93
          Gene Symbol: Raet1d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008758633.1 XP_008756855.1 2 - 3 656 93
          Gene Symbol: Raet1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013063.1 NP_001013081.1 540 93
          GenBank mRNA BC079076.1 - 540 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rrlt
          Click here to find siRNAs for Gene Raet1d
          Click here to find siRNAs for Gene Raet1l

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell mediated cytotoxicity
          antigen processing and presentation of peptide antigen via MHC class I
          positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
          natural killer cell mediated cytotoxicity
          positive regulation of natural killer cell mediated cytotoxicity
          peptide antigen binding
          natural killer cell lectin-like receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P080593 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032614 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P327121 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P080593 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P327121 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P327121 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P080593 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032614 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P327121 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P080593 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10716595 - -
          Affymetrix Rat Gene 1.0 ST Array 10716611 - -
          Affymetrix Rat Gene 1.0 ST Array 10716627 - -
          Affymetrix Rat Gene 1.0 ST Array 10716620 - -
          Affymetrix Rat Gene 1.0 ST Array 10701682 - -
          Affymetrix Rat Gene 1.0 ST Array 10716625 - -
          Affymetrix Rat Gene 1.0 ST Array 10716614 - -
          Affymetrix Rat Gene 1.0 ST Array 10701683 - -
          Affymetrix Rat Gene 1.0 ST Array 10716593 - -
          Affymetrix Rat Gene 1.0 ST Array 10716555 - -
          Affymetrix Rat Gene 1.0 ST Array 10716621 - -
          Affymetrix Rat Gene 1.0 ST Array 10716588 - -
          Affymetrix Rat Gene 1.0 ST Array 10716553 - -
          Affymetrix Rat Gene 1.0 ST Array 10716628 - -
          Affymetrix Rat Gene 1.0 ST Array 10716617 - -
          Affymetrix Rat Gene 1.0 ST Array 10716589 - -
          Affymetrix Rat Gene 1.0 ST Array 10701681 - -
          Affymetrix Rat Gene 1.0 ST Array 10716590 - -
          Affymetrix Rat Gene 1.0 ST Array 10716594 - -
          Affymetrix Rat Gene 1.0 ST Array 10716554 - -
          Affymetrix Rat Gene 1.0 ST Array 10716619 - -
          Affymetrix Rat Gene 1.0 ST Array 10716613 - -
          Affymetrix Rat Gene 1.0 ST Array 10716610 - -
          Affymetrix Rat Gene 1.0 ST Array 10716618 - -
          Affymetrix Rat Gene 1.0 ST Array 10716587 - -
          Affymetrix Rat Gene 1.0 ST Array 10716626 - -
          Affymetrix Rat Gene 1.1 ST Array 10716609 - -
          Affymetrix Rat Gene 1.1 ST Array 10716616 - -
          Affymetrix Rat Gene 1.1 ST Array 10716624 - -
          Affymetrix Rat Gene 1.1 ST Array 10716552 - -
          Affymetrix Rat Gene 1.1 ST Array 10716592 - -
          Affymetrix Rat Clariom S Assay PSR0100032147.rn.1 TC0100003938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032146.rn.1 TC0100003938.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100032155.rn.1 TC0100003938.rn.1 RTA TC
          Array Name Probe IDs Targets Target Type
          RatRef-12 Expression BeadChip ILMN_64782 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0