Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04223779_g1
          See other CESL1 GE Assays ›
          Gene Symbol:
          Cesl1 Ces1f
          Gene Name:
          carboxylesterase-like 1
          carboxylesterase 1F
          Gene Aliases:
          -
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.19: 15081117 - 15126629 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn04223779_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cesl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024365.1 NP_001019536.1 1 56 82
          GenBank mRNA BC079129.1 - 1 56 82
          Gene Symbol: Ces1f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_008772260.1 XP_008770482.1 1 97 82

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC079129.1, NM_001024365.1, XM_008772260.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cesl1
          Click here to find siRNAs for Gene Ces1f

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          short-chain fatty acid catabolic process
          carboxylic ester hydrolase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74832 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65543 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78592 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005108 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005108 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005108 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P470627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95488 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93264 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69777 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17567 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53030 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49420 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44723 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79851 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806204 - -
          Affymetrix Rat Gene 1.0 ST Array 10930683 - -
          Affymetrix Rat Gene 1.0 ST Array 10930677 - -
          Affymetrix Rat Gene 1.0 ST Array 10930684 - -
          Affymetrix Rat Gene 1.0 ST Array 10930676 - -
          Affymetrix Rat Gene 1.0 ST Array 10930681 - -
          Affymetrix Rat Gene 1.0 ST Array 10930680 - -
          Affymetrix Rat Gene 1.0 ST Array 10930682 - -
          Affymetrix Rat Gene 1.0 ST Array 10930679 - -
          Affymetrix Rat Gene 1.0 ST Array 10806209 - -
          Affymetrix Rat Gene 1.0 ST Array 10930678 - -
          Affymetrix Rat Gene 1.1 ST Array 10806200 - -
          Affymetrix Rat Gene 1.1 ST Array 10930675 - -
          Rat Genome 230 2.0 Array 1370352_PM_at - -
          Rat Genome 230 2.0 Array 1370352_at - -
          Affymetrix HT Rat Focus Array 1370352_PM_at - -
          Affymetrix HT Rat Focus Array 1370352_at - -
          Affymetrix Rat Clariom S Assay PSR1900001263.rn.1 TC1900000151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001266.rn.1 TC1900000151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001267.rn.1 TC1900000151.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001268.rn.1 TC1900000151.rn.1 RTA TC
          Rat Expression Set 230 1370352_PM_at - -
          Rat Expression Set 230 1370352_at - -
          Affymetrix Rat Gene 1.0 ST Array 10930675 - -
          Affymetrix Rat Gene 1.0 ST Array 10806200 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21932409 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix