Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn04280217_m1
          See other ETV3L GE Assays ›
          Gene Symbol:
          Etv3l
          Gene Name:
          ets variant 3-like
          Gene Aliases:
          RGD1562620
          Chromosome Location:
          Chr.2: 186899061 - 186911957 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn04280217_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Etv3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100992.2 NP_001094462.2 5 - 6 828 87
          GenBank mRNA BC127525.1 - 5 - 6 828 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001515.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          cell differentiation
          DNA-binding transcription factor activity, RNA polymerase II-specific
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49196 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75569 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067831 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067831 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816305 - -
          Affymetrix Rat Gene 1.0 ST Array 10816308 - -
          Affymetrix Rat Gene 1.0 ST Array 10816307 - -
          Affymetrix Rat Gene 1.0 ST Array 10816306 - -
          Affymetrix Rat Gene 1.0 ST Array 10816304 - -
          Affymetrix Rat Gene 1.1 ST Array 10816303 - -
          Affymetrix Rat Clariom S Assay PSR0200009323.rn.1 TC0200001515.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009324.rn.1 TC0200001515.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200009325.rn.1 TC0200001515.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10816303 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368893 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc