Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI182371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243102.1 | NP_001230031.1 | 2 | 211 | ||
NM_001243103.1 | NP_001230032.1 | 2 | 208 | |||
NM_178885.4 | NP_849216.2 | 2 | 208 | |||
GenBank mRNA | AI046650.1 | - | 2 | 211 | ||
AI287110.1 | - | 2 | 159 | |||
AK050126.1 | - | 2 | 212 | |||
AK050413.1 | - | 2 | 183 | |||
BC019541.1 | - | 2 | 166 | |||
BC037995.1 | - | 2 | 188 | |||
BC094504.1 | - | 2 | 196 | 5100182 | ||
BC115979.1 | - | 2 | 183 | 40104352 | ||
BC115986.1 | - | 2 | 183 | 40104349 |