Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcorl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163073.1 | NP_001156545.1 | 4 | 391 | ||
NM_172153.3 | NP_742165.1 | 4 | 404 | |||
NM_178142.5 | NP_835278.2 | 4 | 391 | |||
GenBank mRNA | AB076078.1 | - | 4 | 403 | ||
AK041987.1 | - | 4 | 350 | |||
AK042567.1 | - | 4 | 405 | |||
AK043498.1 | - | 4 | 349 | |||
AK043845.1 | - | 4 | 359 | |||
AK083689.1 | - | 4 | 353 | |||
AK154398.1 | - | 4 | 352 | |||
AK155421.1 | - | 4 | 352 | |||
BC066151.1 | - | 4 | 321 | |||
BC138563.1 | - | 4 | 391 | 8861583 |