Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cypt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025738.2 | NP_080014.2 | 1 | 104 | ||
GenBank mRNA | AF463502.1 | - | 1 | 85 | ||
AK006892.1 | - | 1 | 107 | |||
AK016467.1 | - | 1 | 106 | |||
AM040451.1 | - | 1 | 98 | |||
AM040454.1 | - | 1 | 98 | |||
AM040455.1 | - | 1 | 98 | |||
BC048455.1 | - | 1 | 103 | |||
BC061079.1 | - | 1 | 103 | 6773161 | ||
BU961231.1 | - | 1 | 106 |
Gene Symbol: Cypt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039943.2 | NP_001035032.1 | 1 | 98 | ||
GenBank mRNA | AM040456.1 | - | 1 | 98 |
Gene Symbol: Cypt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039941.2 | NP_001035030.1 | 1 | 98 | ||
GenBank mRNA | AM040457.1 | - | 1 | 98 |